Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FKRP
Entrez Gene ID
79147
Gene Name
fukutin related protein
Gene Aliases
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5
Chromosome Location
Chr.19: 46746015 - 46758575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039885.2 NP_001034974.1 2 - 3 108 83
NM_024301.4 NP_077277.1 2 - 3 108 83
XM_005259247.2 XP_005259304.1 3 - 4 169 83
XM_005259248.2 XP_005259305.1 3 - 4 211 83
XM_005259249.4 XP_005259306.1 3 - 4 153 83
XM_017027297.1 XP_016882786.1 2 - 3 108 83
GenBank mRNA AJ314847.1 - 2 - 3 96 83
AK022638.1 - 2 - 3 96 83
AK095497.1 - 2 - 3 97 83
AK291282.1 - 2 - 3 96 83
DA123579.1 - 2 - 3 108 83
DB090073.1 - 2 - 3 88 83

View Full Product Details