The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193522.1 | NP_001180451.1 | 14 - 15 | 2737 | 100 |
NM_001193523.1 | NP_001180452.1 | 14 - 15 | 2850 | 100 | |
NM_001193524.1 | NP_001180453.1 | 14 - 15 | 2763 | 100 | |
NM_024519.3 | NP_078795.2 | 14 - 15 | 2728 | 100 | |
XM_011523321.1 | XP_011521623.1 | 14 - 15 | 3542 | 100 | |
XM_011523322.1 | XP_011521624.1 | 14 - 15 | 2737 | 100 | |
XM_011523324.2 | XP_011521626.1 | 14 - 15 | 2797 | 100 | |
XM_011523325.1 | XP_011521627.1 | 14 - 15 | 2752 | 100 | |
XM_011523326.1 | XP_011521628.1 | 14 - 15 | 2951 | 100 | |
XM_017023663.1 | XP_016879152.1 | 14 - 15 | 2734 | 100 | |
XM_017023664.1 | XP_016879153.1 | 13 - 14 | 2623 | 100 | |
GenBank mRNA | AB067517.1 | - | 13 - 14 | 2583 | 100 |
AK023787.1 | - | 9 - 10 | 2220 | 100 | |
AK127792.1 | - | 14 - 15 | 2737 | 100 | |
AK293748.1 | - | 14 - 15 | 2760 | 100 | |
AK295677.1 | - | 14 - 15 | 2694 | 100 | |
AK302868.1 | - | 4 - 5 | 1884 | 100 | |
AL834312.1 | - | 4 - 5 | 1793 | 100 | |
BC054512.1 | - | 4 - 5 | 1770 | 100 | |
BC098587.1 | - | 8 - 9 | 2165 | 100 |