The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024523.5 | NP_078799.2 | 1 - 2 | 1446 | 76 |
GenBank mRNA | AF525417.1 | - | 1 - 2 | 1221 | 76 |
AK025688.1 | - | 1 - 2 | 105 | 76 | |
AK092878.1 | - | 1 - 2 | 902 | 76 | |
AK315472.1 | - | 1 - 2 | 1297 | 76 | |
AL832833.1 | - | 1 - 2 | 108 | 76 | |
BC008902.2 | - | 1 - 2 | 228 | 76 | |
BC014100.2 | - | 1 - 2 | 1446 | 76 | |
BC078665.1 | - | 1 - 2 | 1398 | 76 |