Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44801996-44802490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP8
Entrez Gene ID
23779
Gene Name
Rho GTPase activating protein 8
Gene Aliases
BPGAP1, PP610
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44752558-44862784 on Build GRCh38
Transcripts
NM_001017526.1 NM_001198726.1 NM_181335.2 NM_181334.5

Gene Symbol
PRR5-ARHGAP8
Entrez Gene ID
553158
Gene Name
PRR5-ARHGAP8 readthrough
Gene Aliases
-
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44702198-44862784 on Build GRCh38
Transcripts
NM_001017526.1 NM_001198726.1 NM_181335.2 NM_181334.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243033 c.108C>T p.V36V Chr.22: 44802105-44802105 on Build GRCh38
726888 c.237G>T p.M79I Chr.22: 44802126-44802126 on Build GRCh38
3912952 c.104T>G p.V35G Chr.22: 44802101-44802101 on Build GRCh38
226857 c.137C>T p.S46F Chr.22: 44802134-44802134 on Build GRCh38
7267364 c.101G>A p.R34H Chr.22: 44802098-44802098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753273314 Chr.22: 44802079-44802079 on Build GRCh38
rs910003444 Chr.22: 44802237-44802237 on Build GRCh38
rs761105507 Chr.22: 44802205-44802205 on Build GRCh38
rs144675105 Chr.22: 44802293-44802293 on Build GRCh38
rs199964194 Chr.22: 44802120-44802120 on Build GRCh38
rs372332457 Chr.22: 44802040-44802040 on Build GRCh38
rs755606773 Chr.22: 44802355-44802355 on Build GRCh38
rs772723134 Chr.22: 44802171-44802171 on Build GRCh38
rs779829700 Chr.22: 44802097-44802097 on Build GRCh38
rs1013474508 Chr.22: 44802166-44802166 on Build GRCh38
rs891413454 Chr.22: 44802394-44802394 on Build GRCh38
rs982895883 Chr.22: 44802021-44802021 on Build GRCh38
rs78610026 Chr.22: 44802226-44802226 on Build GRCh38
rs778590600 Chr.22: 44802318-44802318 on Build GRCh38
rs769647517 Chr.22: 44802164-44802164 on Build GRCh38
rs781760854 Chr.22: 44802458-44802458 on Build GRCh38
rs754868850 Chr.22: 44802190-44802212 on Build GRCh38
rs368156067 Chr.22: 44802128-44802128 on Build GRCh38
rs570715679 Chr.22: 44802155-44802155 on Build GRCh38
rs753869236 Chr.22: 44802138-44802138 on Build GRCh38
rs139470300 Chr.22: 44802132-44802132 on Build GRCh38
rs144157668 Chr.22: 44802150-44802150 on Build GRCh38
rs914098263 Chr.22: 44802319-44802319 on Build GRCh38
rs949078534 Chr.22: 44802088-44802088 on Build GRCh38
rs569774676 Chr.22: 44802181-44802182 on Build GRCh38
rs773965825 Chr.22: 44802181-44802181 on Build GRCh38
rs776223636 Chr.22: 44802031-44802031 on Build GRCh38
rs757441997 Chr.22: 44802139-44802139 on Build GRCh38
rs1008472553 Chr.22: 44802410-44802410 on Build GRCh38
rs878984382 Chr.22: 44802148-44802148 on Build GRCh38
rs369071557 Chr.22: 44802250-44802250 on Build GRCh38
rs764520838 Chr.22: 44802211-44802211 on Build GRCh38
rs572175871 Chr.22: 44802450-44802450 on Build GRCh38
rs936428309 Chr.22: 44802309-44802309 on Build GRCh38
rs995444608 Chr.22: 44802467-44802467 on Build GRCh38
rs777764946 Chr.22: 44802082-44802082 on Build GRCh38
rs754784555 Chr.22: 44802152-44802152 on Build GRCh38
rs371309902 Chr.22: 44802131-44802131 on Build GRCh38
rs764904016 Chr.22: 44802042-44802042 on Build GRCh38
rs769321323 Chr.22: 44802108-44802108 on Build GRCh38
rs1002341754 Chr.22: 44802466-44802466 on Build GRCh38
rs934116151 Chr.22: 44802302-44802302 on Build GRCh38
rs775865926 Chr.22: 44802192-44802192 on Build GRCh38
rs951417136 Chr.22: 44802186-44802186 on Build GRCh38
rs201465628 Chr.22: 44802101-44802101 on Build GRCh38
rs41278883 Chr.22: 44802154-44802154 on Build GRCh38
rs949329006 Chr.22: 44802253-44802253 on Build GRCh38
rs975604328 Chr.22: 44802270-44802270 on Build GRCh38
rs867090648 Chr.22: 44802411-44802411 on Build GRCh38
rs2239813 Chr.22: 44802129-44802129 on Build GRCh38
rs751878132 Chr.22: 44802067-44802067 on Build GRCh38
rs746159688 Chr.22: 44802147-44802147 on Build GRCh38
rs772886283 Chr.22: 44802044-44802044 on Build GRCh38
rs1046947150 Chr.22: 44802047-44802047 on Build GRCh38
rs866890566 Chr.22: 44802086-44802086 on Build GRCh38
rs539384500 Chr.22: 44802183-44802183 on Build GRCh38
rs181719747 Chr.22: 44802122-44802122 on Build GRCh38
rs974115385 Chr.22: 44802172-44802172 on Build GRCh38
rs555461655 Chr.22: 44802296-44802296 on Build GRCh38
rs57415948 Chr.22: 44802019-44802019 on Build GRCh38
rs768629591 Chr.22: 44802099-44802099 on Build GRCh38
rs570717841 Chr.22: 44802416-44802416 on Build GRCh38
rs772380059 Chr.22: 44802184-44802184 on Build GRCh38
rs12159794 Chr.22: 44802071-44802071 on Build GRCh38
rs1041852693 Chr.22: 44802388-44802388 on Build GRCh38
rs569752689 Chr.22: 44802085-44802085 on Build GRCh38
rs770544788 Chr.22: 44802178-44802178 on Build GRCh38
rs774113480 Chr.22: 44802124-44802124 on Build GRCh38
rs963106827 Chr.22: 44802247-44802247 on Build GRCh38
rs748737339 Chr.22: 44802174-44802174 on Build GRCh38
rs576078872 Chr.22: 44802246-44802246 on Build GRCh38
rs766387703 Chr.22: 44802164-44802167 on Build GRCh38
rs748132766 Chr.22: 44802107-44802107 on Build GRCh38
rs768002077 Chr.22: 44802075-44802075 on Build GRCh38
rs1027034170 Chr.22: 44802468-44802468 on Build GRCh38
rs375054113 Chr.22: 44802093-44802093 on Build GRCh38
rs756156013 Chr.22: 44802080-44802080 on Build GRCh38
rs980879140 Chr.22: 44802283-44802283 on Build GRCh38
rs749933320 Chr.22: 44802337-44802337 on Build GRCh38
rs201611417 Chr.22: 44802121-44802121 on Build GRCh38
rs749479656 Chr.22: 44802083-44802083 on Build GRCh38
rs368399115 Chr.22: 44802098-44802098 on Build GRCh38
rs778988324 Chr.22: 44802141-44802141 on Build GRCh38
rs766714509 Chr.22: 44802056-44802056 on Build GRCh38
rs755422184 Chr.22: 44802073-44802073 on Build GRCh38
rs1043776060 Chr.22: 44802419-44802419 on Build GRCh38

View Full Product Details