Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44845241-44845751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP8
Entrez Gene ID
23779
Gene Name
Rho GTPase activating protein 8
Gene Aliases
BPGAP1, PP610
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44752558-44862784 on Build GRCh38
Transcripts
NM_001017526.1 NM_001198726.1 NM_181335.2 NM_181334.5

Gene Symbol
PRR5-ARHGAP8
Entrez Gene ID
553158
Gene Name
PRR5-ARHGAP8 readthrough
Gene Aliases
-
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44702198-44862784 on Build GRCh38
Transcripts
NM_001017526.1 NM_001198726.1 NM_181335.2 NM_181334.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563638 c.709G>T p.G237C Chr.22: 44845288-44845288 on Build GRCh38
5393815 c.738C>T p.P246P Chr.22: 44845302-44845302 on Build GRCh38
5909863 c.721C>T p.P241S Chr.22: 44845300-44845300 on Build GRCh38
1308253 c.753G>C p.L251L Chr.22: 44845332-44845332 on Build GRCh38
1632643 c.773A>G p.E258G Chr.22: 44845337-44845337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914983158 Chr.22: 44845639-44845639 on Build GRCh38
rs557061581 Chr.22: 44845622-44845622 on Build GRCh38
rs765957967 Chr.22: 44845498-44845498 on Build GRCh38
rs1007905174 Chr.22: 44845597-44845597 on Build GRCh38
rs901506412 Chr.22: 44845670-44845670 on Build GRCh38
rs776407512 Chr.22: 44845372-44845372 on Build GRCh38
rs965787751 Chr.22: 44845453-44845453 on Build GRCh38
rs757767644 Chr.22: 44845644-44845644 on Build GRCh38
rs200724454 Chr.22: 44845329-44845329 on Build GRCh38
rs759087263 Chr.22: 44845319-44845319 on Build GRCh38
rs765201276 Chr.22: 44845356-44845357 on Build GRCh38
rs982407773 Chr.22: 44845575-44845575 on Build GRCh38
rs916102504 Chr.22: 44845434-44845434 on Build GRCh38
rs16992952 Chr.22: 44845423-44845423 on Build GRCh38
rs373419119 Chr.22: 44845373-44845373 on Build GRCh38
rs761251880 Chr.22: 44845610-44845610 on Build GRCh38
rs915252348 Chr.22: 44845357-44845357 on Build GRCh38
rs757713884 Chr.22: 44845290-44845290 on Build GRCh38
rs997246655 Chr.22: 44845701-44845701 on Build GRCh38
rs970345743 Chr.22: 44845555-44845555 on Build GRCh38
rs996960398 Chr.22: 44845495-44845495 on Build GRCh38
rs566133332 Chr.22: 44845413-44845413 on Build GRCh38
rs540915857 Chr.22: 44845291-44845291 on Build GRCh38
rs986321587 Chr.22: 44845328-44845328 on Build GRCh38
rs956100671 Chr.22: 44845713-44845713 on Build GRCh38
rs1031380795 Chr.22: 44845704-44845704 on Build GRCh38
rs398101786 Chr.22: 44845553-44845554 on Build GRCh38
rs573007444 Chr.22: 44845663-44845663 on Build GRCh38
rs370256316 Chr.22: 44845363-44845363 on Build GRCh38
rs929480380 Chr.22: 44845614-44845614 on Build GRCh38
rs781240864 Chr.22: 44845387-44845387 on Build GRCh38
rs762584319 Chr.22: 44845490-44845490 on Build GRCh38
rs9626576 Chr.22: 44845562-44845562 on Build GRCh38
rs113660416 Chr.22: 44845675-44845675 on Build GRCh38
rs145431642 Chr.22: 44845325-44845325 on Build GRCh38
rs1007233300 Chr.22: 44845439-44845439 on Build GRCh38
rs201724810 Chr.22: 44845336-44845336 on Build GRCh38
rs868157781 Chr.22: 44845471-44845471 on Build GRCh38
rs926580987 Chr.22: 44845576-44845576 on Build GRCh38
rs374689628 Chr.22: 44845425-44845425 on Build GRCh38
rs994178597 Chr.22: 44845429-44845429 on Build GRCh38
rs888535628 Chr.22: 44845705-44845705 on Build GRCh38
rs990506394 Chr.22: 44845428-44845428 on Build GRCh38
rs961327141 Chr.22: 44845559-44845559 on Build GRCh38
rs755157118 Chr.22: 44845347-44845347 on Build GRCh38
rs776188902 Chr.22: 44845529-44845529 on Build GRCh38
rs533465387 Chr.22: 44845638-44845638 on Build GRCh38
rs574005292 Chr.22: 44845645-44845645 on Build GRCh38
rs747821034 Chr.22: 44845368-44845368 on Build GRCh38
rs377592891 Chr.22: 44845355-44845355 on Build GRCh38
rs549109058 Chr.22: 44845395-44845395 on Build GRCh38
rs768183510 Chr.22: 44845370-44845370 on Build GRCh38
rs997195442 Chr.22: 44845454-44845454 on Build GRCh38
rs147582478 Chr.22: 44845299-44845299 on Build GRCh38
rs552007156 Chr.22: 44845437-44845437 on Build GRCh38
rs760350190 Chr.22: 44845364-44845364 on Build GRCh38
rs751651840 Chr.22: 44845384-44845384 on Build GRCh38
rs536342998 Chr.22: 44845654-44845654 on Build GRCh38
rs764437007 Chr.22: 44845722-44845722 on Build GRCh38
rs1056543501 Chr.22: 44845598-44845598 on Build GRCh38
rs754333566 Chr.22: 44845266-44845266 on Build GRCh38
rs766377766 Chr.22: 44845382-44845382 on Build GRCh38
rs67893074 Chr.22: 44845552-44845553 on Build GRCh38
rs878870264 Chr.22: 44845520-44845520 on Build GRCh38
rs1005381855 Chr.22: 44845546-44845546 on Build GRCh38
rs747190851 Chr.22: 44845341-44845341 on Build GRCh38
rs1041066282 Chr.22: 44845469-44845469 on Build GRCh38
rs571941037 Chr.22: 44845497-44845497 on Build GRCh38
rs563957611 Chr.22: 44845669-44845669 on Build GRCh38
rs145150063 Chr.22: 44845294-44845294 on Build GRCh38
rs560799512 Chr.22: 44845460-44845460 on Build GRCh38
rs563332820 Chr.22: 44845349-44845349 on Build GRCh38
rs146372974 Chr.22: 44845326-44845326 on Build GRCh38
rs771669473 Chr.22: 44845356-44845356 on Build GRCh38
rs1017182397 Chr.22: 44845451-44845451 on Build GRCh38
rs770800221 Chr.22: 44845351-44845351 on Build GRCh38
rs141993522 Chr.22: 44845304-44845304 on Build GRCh38
rs980813275 Chr.22: 44845571-44845571 on Build GRCh38
rs187729236 Chr.22: 44845522-44845522 on Build GRCh38
rs763481058 Chr.22: 44845377-44845377 on Build GRCh38
rs763590508 Chr.22: 44845458-44845458 on Build GRCh38
rs749023096 Chr.22: 44845350-44845350 on Build GRCh38
rs866989460 Chr.22: 44845611-44845611 on Build GRCh38
rs750964191 Chr.22: 44845334-44845334 on Build GRCh38
rs1049878885 Chr.22: 44845697-44845697 on Build GRCh38
rs1056658915 Chr.22: 44845404-44845404 on Build GRCh38
rs775147571 Chr.22: 44845358-44845358 on Build GRCh38
rs773117999 Chr.22: 44845305-44845305 on Build GRCh38
rs772909625 Chr.22: 44845303-44845303 on Build GRCh38
rs772680966 Chr.22: 44845477-44845477 on Build GRCh38
rs12159141 Chr.22: 44845656-44845656 on Build GRCh38
rs774415242 Chr.22: 44845317-44845317 on Build GRCh38
rs139301758 Chr.22: 44845666-44845666 on Build GRCh38
rs759502919 Chr.22: 44845332-44845333 on Build GRCh38
rs1006017207 Chr.22: 44845708-44845708 on Build GRCh38
rs755143334 Chr.22: 44845385-44845385 on Build GRCh38
rs1049313927 Chr.22: 44845435-44845435 on Build GRCh38
rs532804701 Chr.22: 44845720-44845720 on Build GRCh38
rs772206596 Chr.22: 44845278-44845278 on Build GRCh38

View Full Product Details