The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024607.3 | NP_078883.2 | 1 - 2 | 151 | 143 |
GenBank mRNA | AK024067.1 | - | 1 - 2 | 137 | 143 |
BC043388.1 | - | 1 - 2 | 84 | 143 | |
BQ431880.1 | - | 1 - 2 | 83 | 143 | |
DA100998.1 | - | 1 - 2 | 151 | 143 |