The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039706.2 | NP_001034795.2 | 15 - 16 | 2030 | 147 |
NM_001160138.1 | NP_001153610.1 | 15 - 16 | 1976 | 147 | |
XM_011516577.2 | XP_011514879.1 | 15 - 16 | 2009 | 147 | |
XM_011516578.2 | XP_011514880.1 | 15 - 16 | 2009 | 147 | |
XM_017012624.1 | XP_016868113.1 | 15 - 16 | 1955 | 147 | |
XM_017012625.1 | XP_016868114.1 | 15 - 16 | 2009 | 147 | |
XM_017012626.1 | XP_016868115.1 | 15 - 16 | 2009 | 147 | |
XM_017012627.1 | XP_016868116.1 | 15 - 16 | 2009 | 147 | |
XM_017012628.1 | XP_016868117.1 | 15 - 16 | 2009 | 147 | |
XM_017012629.1 | XP_016868118.1 | 14 - 15 | 1899 | 147 | |
XM_017012630.1 | XP_016868119.1 | 15 - 16 | 2009 | 147 | |
XM_017012631.1 | XP_016868120.1 | 13 - 14 | 1557 | 147 | |
XM_017012632.1 | XP_016868121.1 | 15 - 16 | 2009 | 147 | |
XM_017012633.1 | XP_016868122.1 | 13 - 14 | 4586 | 147 | |
XM_017012634.1 | XP_016868123.1 | 13 - 14 | 1724 | 147 | |
XM_017012635.1 | XP_016868124.1 | 11 - 12 | 1420 | 147 | |
XM_017012636.1 | XP_016868125.1 | 10 - 11 | 1361 | 147 | |
XM_017012637.1 | XP_016868126.1 | 11 - 12 | 4526 | 147 | |
XM_017012638.1 | XP_016868127.1 | 11 - 12 | 1367 | 147 | |
XM_017012639.1 | XP_016868128.1 | 11 - 12 | 1402 | 147 | |
XM_017012640.1 | XP_016868129.1 | 8 - 9 | 1006 | 147 | |
XM_017012641.1 | XP_016868130.1 | 15 - 16 | 2009 | 147 | |
GenBank mRNA | AK024715.1 | - | 4 - 5 | 528 | 147 |
AK093991.1 | - | 13 - 14 | 1812 | 147 | |
AK302636.1 | - | 15 - 16 | 1935 | 147 | |
AK303034.1 | - | 15 - 16 | 2009 | 147 | |
AL833446.1 | - | 9 - 10 | 1223 | 147 | |
BC036351.1 | - | 16 - 17 | 2194 | 147 | |
BC132771.1 | - | 9 - 10 | 1252 | 147 | |
BC140742.1 | - | 15 - 16 | 1916 | 147 | |
BC141834.1 | - | 15 - 16 | 1976 | 147 | |
BC142655.1 | - | 15 - 16 | 1976 | 147 |