The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317902.1 | NP_001304831.1 | 8 - 9 | 2321 | 64 |
NM_024831.7 | NP_079107.6 | 9 - 10 | 2386 | 64 | |
XM_005251328.2 | XP_005251385.1 | 8 - 9 | 2101 | 64 | |
XM_006716485.2 | XP_006716548.1 | 9 - 10 | 2172 | 64 | |
XM_017014004.1 | XP_016869493.1 | 9 - 10 | 2172 | 64 | |
GenBank mRNA | AF286340.1 | - | 9 - 10 | 2476 | 64 |
AF432215.1 | - | 9 - 10 | 2134 | 64 | |
AK026648.1 | - | 1 - 2 | 163 | 64 | |
AK055178.1 | - | 6 - 7 | 1260 | 64 | |
AK314693.1 | - | 9 - 10 | 2160 | 64 | |
AL832426.1 | - | 3 - 4 | 642 | 64 | |
AY028423.1 | - | 9 - 10 | 2167 | 64 | |
AY534911.1 | - | 6 - 7 | 1176 | 64 | |
BC011999.1 | - | 2 - 3 | 280 | 64 | |
BX648548.1 | - | 8 - 9 | 2084 | 64 |