Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94172744-94173253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP29
Entrez Gene ID
9411
Gene Name
Rho GTPase activating protein 29
Gene Aliases
PARG1
UniGene
Hs.483238
Gene Chromosome Location
Chr.1: 94168907-94275068 on Build GRCh38
Transcripts
NM_004815.3 XM_006711048.3 XM_011542438.2 XM_011542439.2 XM_011542440.2

SNP Detail

SNP ID Chromosome Location
rs186345482 Chr.1: 94172817-94172817 on Build GRCh38
rs545976707 Chr.1: 94173014-94173014 on Build GRCh38
rs1006995253 Chr.1: 94172954-94172954 on Build GRCh38
rs950661268 Chr.1: 94173073-94173073 on Build GRCh38
rs976016763 Chr.1: 94172778-94172778 on Build GRCh38
rs761231464 Chr.1: 94173168-94173172 on Build GRCh38
rs1016138649 Chr.1: 94173043-94173043 on Build GRCh38
rs189443233 Chr.1: 94172861-94172861 on Build GRCh38
rs949586795 Chr.1: 94173156-94173156 on Build GRCh38
rs544506940 Chr.1: 94173021-94173021 on Build GRCh38
rs370309447 Chr.1: 94172794-94172794 on Build GRCh38
rs908890166 Chr.1: 94173079-94173079 on Build GRCh38
rs946000139 Chr.1: 94173212-94173212 on Build GRCh38
rs17111192 Chr.1: 94172891-94172891 on Build GRCh38
rs1018349480 Chr.1: 94172981-94172981 on Build GRCh38
rs763367318 Chr.1: 94172909-94172909 on Build GRCh38
rs11196 Chr.1: 94172883-94172883 on Build GRCh38
rs1002985679 Chr.1: 94173004-94173004 on Build GRCh38
rs544219139 Chr.1: 94173195-94173195 on Build GRCh38
rs941793252 Chr.1: 94172810-94172810 on Build GRCh38
rs774632980 Chr.1: 94173129-94173129 on Build GRCh38
rs201360137 Chr.1: 94172804-94172804 on Build GRCh38
rs983734336 Chr.1: 94173076-94173076 on Build GRCh38
rs542107032 Chr.1: 94173108-94173108 on Build GRCh38
rs572997888 Chr.1: 94173077-94173077 on Build GRCh38
rs907275989 Chr.1: 94172972-94172972 on Build GRCh38
rs139595133 Chr.1: 94173172-94173172 on Build GRCh38
rs567010882 Chr.1: 94172822-94172822 on Build GRCh38
rs139214292 Chr.1: 94172869-94172869 on Build GRCh38
rs181497124 Chr.1: 94173202-94173202 on Build GRCh38
rs975902115 Chr.1: 94173049-94173049 on Build GRCh38
rs147874683 Chr.1: 94172809-94172809 on Build GRCh38
rs893766894 Chr.1: 94172935-94172935 on Build GRCh38
rs902638591 Chr.1: 94173186-94173186 on Build GRCh38
rs971717575 Chr.1: 94173112-94173112 on Build GRCh38
rs964565578 Chr.1: 94173042-94173042 on Build GRCh38
rs573795134 Chr.1: 94173225-94173225 on Build GRCh38
rs146489766 Chr.1: 94173028-94173028 on Build GRCh38
rs766727797 Chr.1: 94173092-94173092 on Build GRCh38
rs577036340 Chr.1: 94172956-94172956 on Build GRCh38
rs1048866 Chr.1: 94173155-94173155 on Build GRCh38
rs1030699567 Chr.1: 94173059-94173059 on Build GRCh38
rs550642749 Chr.1: 94172774-94172774 on Build GRCh38
rs1046658438 Chr.1: 94172882-94172882 on Build GRCh38
rs923167082 Chr.1: 94172787-94172787 on Build GRCh38
rs372335202 Chr.1: 94172910-94172910 on Build GRCh38
rs922969325 Chr.1: 94173127-94173127 on Build GRCh38
rs760665893 Chr.1: 94172859-94172859 on Build GRCh38
rs1034596507 Chr.1: 94173019-94173019 on Build GRCh38
rs942943426 Chr.1: 94172852-94172852 on Build GRCh38
rs114512647 Chr.1: 94172792-94172792 on Build GRCh38
rs1041723835 Chr.1: 94173223-94173223 on Build GRCh38

View Full Product Details