Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94474864-94475374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD3
Entrez Gene ID
5825
Gene Name
ATP binding cassette subfamily D member 3
Gene Aliases
ABC43, CBAS5, PMP70, PXMP1, ZWS2
UniGene
Hs.700576
Gene Chromosome Location
Chr.1: 94385106-94518663 on Build GRCh38
Transcripts
NM_001122674.1 NM_002858.3 XM_006710802.2 XM_017001952.1 XM_017001953.1 XM_017001954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423125 c.463C>T p.R155* Chr.1: 94475200-94475200 on Build GRCh38
1584734 c.410C>T p.S137F Chr.1: 94475147-94475147 on Build GRCh38
5754535 c.470G>T p.R157M Chr.1: 94475207-94475207 on Build GRCh38
6019510 c.476C>G p.T159S Chr.1: 94475213-94475213 on Build GRCh38
6019509 c.476C>G p.T159S Chr.1: 94475213-94475213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201088281 Chr.1: 94475207-94475207 on Build GRCh38
rs1002826241 Chr.1: 94475006-94475007 on Build GRCh38
rs1041151639 Chr.1: 94474896-94474896 on Build GRCh38
rs540569898 Chr.1: 94475157-94475157 on Build GRCh38
rs1037185416 Chr.1: 94475061-94475061 on Build GRCh38
rs776609408 Chr.1: 94475219-94475219 on Build GRCh38
rs777761655 Chr.1: 94475184-94475184 on Build GRCh38
rs542205756 Chr.1: 94474952-94474952 on Build GRCh38
rs547113264 Chr.1: 94474897-94474897 on Build GRCh38
rs890944862 Chr.1: 94474974-94474974 on Build GRCh38
rs746977470 Chr.1: 94475190-94475190 on Build GRCh38
rs769203193 Chr.1: 94475280-94475282 on Build GRCh38
rs1023598625 Chr.1: 94474894-94474894 on Build GRCh38
rs1052970106 Chr.1: 94475331-94475331 on Build GRCh38
rs113318251 Chr.1: 94475349-94475349 on Build GRCh38
rs781500412 Chr.1: 94475137-94475137 on Build GRCh38
rs932502390 Chr.1: 94474960-94474960 on Build GRCh38
rs935813316 Chr.1: 94475283-94475283 on Build GRCh38
rs768086676 Chr.1: 94475099-94475099 on Build GRCh38
rs921150471 Chr.1: 94474956-94474956 on Build GRCh38
rs758947202 Chr.1: 94475124-94475125 on Build GRCh38
rs184858284 Chr.1: 94474910-94474910 on Build GRCh38
rs749484549 Chr.1: 94475223-94475223 on Build GRCh38
rs531656312 Chr.1: 94474947-94474947 on Build GRCh38
rs780536006 Chr.1: 94475125-94475128 on Build GRCh38
rs761132918 Chr.1: 94475263-94475263 on Build GRCh38
rs754925541 Chr.1: 94474905-94474905 on Build GRCh38
rs148757295 Chr.1: 94474966-94474966 on Build GRCh38
rs773752722 Chr.1: 94475262-94475262 on Build GRCh38
rs1052939715 Chr.1: 94474951-94474951 on Build GRCh38
rs553271925 Chr.1: 94474912-94474912 on Build GRCh38
rs754664540 Chr.1: 94475144-94475144 on Build GRCh38
rs914619082 Chr.1: 94474937-94474937 on Build GRCh38
rs148319104 Chr.1: 94475175-94475175 on Build GRCh38
rs562278519 Chr.1: 94475107-94475107 on Build GRCh38
rs374064414 Chr.1: 94475255-94475255 on Build GRCh38
rs562872176 Chr.1: 94474911-94474911 on Build GRCh38
rs111707969 Chr.1: 94475179-94475179 on Build GRCh38
rs112436069 Chr.1: 94475285-94475285 on Build GRCh38
rs544746668 Chr.1: 94475100-94475100 on Build GRCh38
rs573151194 Chr.1: 94474931-94474931 on Build GRCh38
rs564509016 Chr.1: 94475115-94475115 on Build GRCh38
rs570653204 Chr.1: 94474909-94474909 on Build GRCh38
rs778354023 Chr.1: 94475151-94475151 on Build GRCh38
rs753464068 Chr.1: 94475143-94475143 on Build GRCh38
rs970322216 Chr.1: 94475193-94475193 on Build GRCh38
rs766697116 Chr.1: 94475267-94475267 on Build GRCh38
rs560793659 Chr.1: 94475225-94475225 on Build GRCh38
rs761962129 Chr.1: 94475233-94475233 on Build GRCh38
rs113623858 Chr.1: 94474955-94474955 on Build GRCh38
rs908555426 Chr.1: 94474891-94474891 on Build GRCh38
rs547746601 Chr.1: 94475337-94475337 on Build GRCh38
rs267598781 Chr.1: 94475199-94475199 on Build GRCh38
rs748203806 Chr.1: 94475303-94475303 on Build GRCh38
rs141432588 Chr.1: 94475232-94475232 on Build GRCh38
rs879377651 Chr.1: 94475064-94475064 on Build GRCh38
rs113004664 Chr.1: 94475046-94475046 on Build GRCh38
rs373460632 Chr.1: 94475119-94475119 on Build GRCh38
rs989296561 Chr.1: 94474914-94474914 on Build GRCh38
rs747752446 Chr.1: 94475266-94475266 on Build GRCh38
rs1033837378 Chr.1: 94475044-94475044 on Build GRCh38

View Full Product Details