The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085437.2 | NP_001078906.2 | 2 - 3 | 899 | 100 |
NM_001282921.1 | NP_001269850.1 | 2 - 3 | 392 | 100 | |
NM_024861.3 | NP_079137.3 | 2 - 3 | 335 | 100 | |
XM_011511877.1 | XP_011510179.1 | 3 - 4 | 891 | 100 | |
GenBank mRNA | AK026324.1 | - | 2 - 3 | 332 | 100 |
AK056601.1 | - | 2 - 3 | 899 | 100 | |
AK096434.1 | - | 2 - 3 | 392 | 100 | |
BC125154.1 | - | 2 - 3 | 323 | 100 | |
BC125155.1 | - | 2 - 3 | 323 | 100 |