Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94982599-94983100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG14
Entrez Gene ID
199857
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CMS15
UniGene
Hs.408927
Gene Chromosome Location
Chr.1: 94981565-95072974 on Build GRCh38
Transcripts
NM_001305242.1 NM_144988.3 NR_131032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1345036 c.641G>A p.R214Q Chr.1: 94983086-94983086 on Build GRCh38
682629 c.640C>A p.R214R Chr.1: 94983087-94983087 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs113133699 Chr.1: 94983076-94983076 on Build GRCh38
rs987329231 Chr.1: 94982745-94982745 on Build GRCh38
rs955691169 Chr.1: 94982988-94982988 on Build GRCh38
rs934354348 Chr.1: 94982674-94982674 on Build GRCh38
rs557052288 Chr.1: 94982933-94982933 on Build GRCh38
rs56812521 Chr.1: 94982722-94982723 on Build GRCh38
rs149024884 Chr.1: 94982844-94982844 on Build GRCh38
rs760367713 Chr.1: 94983069-94983069 on Build GRCh38
rs905733510 Chr.1: 94982744-94982744 on Build GRCh38
rs112318591 Chr.1: 94982875-94982875 on Build GRCh38
rs778761373 Chr.1: 94982891-94982891 on Build GRCh38
rs564250428 Chr.1: 94982729-94982729 on Build GRCh38
rs6694639 Chr.1: 94982698-94982698 on Build GRCh38
rs949825162 Chr.1: 94983006-94983006 on Build GRCh38
rs979844768 Chr.1: 94983013-94983013 on Build GRCh38
rs180812585 Chr.1: 94982868-94982868 on Build GRCh38
rs1040766612 Chr.1: 94983068-94983068 on Build GRCh38
rs369799841 Chr.1: 94982920-94982920 on Build GRCh38
rs112335700 Chr.1: 94983069-94983069 on Build GRCh38
rs913400574 Chr.1: 94982999-94982999 on Build GRCh38
rs1005295750 Chr.1: 94982782-94982782 on Build GRCh38
rs946171388 Chr.1: 94983008-94983008 on Build GRCh38
rs570453160 Chr.1: 94982982-94982982 on Build GRCh38
rs1014756147 Chr.1: 94982761-94982761 on Build GRCh38
rs6682185 Chr.1: 94982640-94982640 on Build GRCh38
rs764623435 Chr.1: 94983059-94983059 on Build GRCh38
rs530640275 Chr.1: 94982666-94982666 on Build GRCh38
rs368855559 Chr.1: 94982699-94982700 on Build GRCh38
rs886409306 Chr.1: 94982774-94982775 on Build GRCh38
rs544253800 Chr.1: 94982671-94982671 on Build GRCh38
rs765319440 Chr.1: 94983037-94983037 on Build GRCh38
rs144953323 Chr.1: 94982736-94982736 on Build GRCh38
rs143040411 Chr.1: 94983063-94983063 on Build GRCh38
rs386367730 Chr.1: 94982700-94982701 on Build GRCh38
rs973348525 Chr.1: 94982840-94982840 on Build GRCh38
rs547996899 Chr.1: 94983005-94983006 on Build GRCh38
rs558518381 Chr.1: 94982843-94982843 on Build GRCh38
rs1029149108 Chr.1: 94982881-94982881 on Build GRCh38
rs759471629 Chr.1: 94983029-94983029 on Build GRCh38
rs566886425 Chr.1: 94982762-94982762 on Build GRCh38
rs763442364 Chr.1: 94983055-94983055 on Build GRCh38
rs1052837357 Chr.1: 94982683-94982683 on Build GRCh38
rs552681984 Chr.1: 94983020-94983020 on Build GRCh38
rs757661309 Chr.1: 94983064-94983064 on Build GRCh38
rs538442637 Chr.1: 94982882-94982882 on Build GRCh38
rs539550497 Chr.1: 94983004-94983004 on Build GRCh38
rs1057266366 Chr.1: 94982901-94982901 on Build GRCh38
rs994694124 Chr.1: 94982760-94982760 on Build GRCh38
rs533507065 Chr.1: 94982733-94982733 on Build GRCh38
rs886374685 Chr.1: 94983050-94983050 on Build GRCh38
rs775600527 Chr.1: 94983041-94983041 on Build GRCh38
rs979705997 Chr.1: 94982651-94982651 on Build GRCh38
rs904196443 Chr.1: 94982641-94982641 on Build GRCh38
rs777079109 Chr.1: 94983033-94983033 on Build GRCh38
rs376803101 Chr.1: 94983031-94983031 on Build GRCh38
rs916976401 Chr.1: 94983006-94983006 on Build GRCh38
rs79206206 Chr.1: 94983022-94983022 on Build GRCh38
rs569443425 Chr.1: 94982881-94982881 on Build GRCh38

View Full Product Details