The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144875.1 | NP_001138347.1 | 3 - 4 | 576 | 85 |
NM_001308235.1 | NP_001295164.1 | 3 - 4 | 768 | 85 | |
NM_001308236.1 | NP_001295165.1 | 3 - 4 | 591 | 85 | |
NM_024872.2 | NP_079148.2 | 3 - 4 | 547 | 85 | |
XM_005265985.3 | XP_005266042.1 | 3 - 4 | 604 | 85 | |
XM_005265987.1 | XP_005266044.1 | 3 - 4 | 604 | 85 | |
XM_011534654.2 | XP_011532956.1 | 3 - 4 | 481 | 85 | |
XM_011534655.2 | XP_011532957.1 | 3 - 4 | 657 | 85 | |
XM_011534656.2 | XP_011532958.1 | 3 - 4 | 458 | 85 | |
GenBank mRNA | AK026223.1 | - | 3 - 4 | 542 | 85 |
AL583057.3 | - | 3 - 4 | 722 | 85 | |
BC004564.2 | - | 3 - 4 | 535 | 85 | |
BC004867.2 | - | 1 - 2 | 643 | 85 | |
BI489608.1 | - | 3 - 4 | 569 | 85 |