Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15667514-15668042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
UniGene
Hs.335239
Gene Chromosome Location
Chr.3: 15667236-15859814 on Build GRCh38
Transcripts
NM_001195098.1 NM_001195099.1 NM_015199.3 XM_011533540.2

SNP Detail

SNP ID Chromosome Location
rs915213012 Chr.3: 15668003-15668003 on Build GRCh38
rs776568083 Chr.3: 15667906-15667908 on Build GRCh38
rs956981399 Chr.3: 15667948-15667948 on Build GRCh38
rs1056819237 Chr.3: 15667666-15667666 on Build GRCh38
rs963691704 Chr.3: 15667774-15667774 on Build GRCh38
rs953529431 Chr.3: 15667839-15667839 on Build GRCh38
rs552430875 Chr.3: 15667746-15667746 on Build GRCh38
rs1029204781 Chr.3: 15667821-15667821 on Build GRCh38
rs538886383 Chr.3: 15667628-15667628 on Build GRCh38
rs546609143 Chr.3: 15667662-15667662 on Build GRCh38
rs1004572045 Chr.3: 15667697-15667697 on Build GRCh38
rs947339199 Chr.3: 15668008-15668008 on Build GRCh38
rs777371422 Chr.3: 15667686-15667686 on Build GRCh38
rs911924590 Chr.3: 15667578-15667578 on Build GRCh38
rs1046952 Chr.3: 15667890-15667890 on Build GRCh38
rs760664712 Chr.3: 15667556-15667557 on Build GRCh38
rs953476647 Chr.3: 15667564-15667564 on Build GRCh38
rs936240769 Chr.3: 15667761-15667761 on Build GRCh38
rs561862573 Chr.3: 15667604-15667604 on Build GRCh38
rs371149477 Chr.3: 15667590-15667591 on Build GRCh38
rs905695203 Chr.3: 15667660-15667660 on Build GRCh38
rs141461081 Chr.3: 15667708-15667709 on Build GRCh38
rs190429199 Chr.3: 15667785-15667785 on Build GRCh38
rs969501751 Chr.3: 15667678-15667678 on Build GRCh38
rs1052834679 Chr.3: 15667615-15667615 on Build GRCh38
rs943357752 Chr.3: 15667722-15667743 on Build GRCh38
rs987551808 Chr.3: 15667577-15667577 on Build GRCh38
rs988182007 Chr.3: 15667987-15667987 on Build GRCh38
rs927171972 Chr.3: 15668021-15668021 on Build GRCh38
rs1053279737 Chr.3: 15667776-15667776 on Build GRCh38
rs900534896 Chr.3: 15667538-15667538 on Build GRCh38
rs1014987250 Chr.3: 15667731-15667731 on Build GRCh38
rs139014065 Chr.3: 15667807-15667807 on Build GRCh38
rs542104945 Chr.3: 15667566-15667566 on Build GRCh38
rs1042948893 Chr.3: 15668012-15668012 on Build GRCh38
rs560166187 Chr.3: 15667682-15667682 on Build GRCh38
rs983600 Chr.3: 15667633-15667633 on Build GRCh38
rs995080795 Chr.3: 15667786-15667786 on Build GRCh38
rs922082735 Chr.3: 15667547-15667547 on Build GRCh38
rs960564558 Chr.3: 15668002-15668002 on Build GRCh38
rs1027987286 Chr.3: 15667579-15667579 on Build GRCh38
rs143039577 Chr.3: 15667833-15667833 on Build GRCh38
rs913430259 Chr.3: 15667800-15667800 on Build GRCh38
rs181747037 Chr.3: 15667880-15667880 on Build GRCh38
rs1014837071 Chr.3: 15667960-15667960 on Build GRCh38
rs951963319 Chr.3: 15667617-15667617 on Build GRCh38
rs533742644 Chr.3: 15667942-15667942 on Build GRCh38
rs11920039 Chr.3: 15667919-15667919 on Build GRCh38
rs771653111 Chr.3: 15667667-15667667 on Build GRCh38
rs996587532 Chr.3: 15667889-15667889 on Build GRCh38
rs913166067 Chr.3: 15667623-15667623 on Build GRCh38
rs1004827537 Chr.3: 15667950-15667950 on Build GRCh38
rs887709312 Chr.3: 15667941-15667941 on Build GRCh38
rs532478030 Chr.3: 15667744-15667744 on Build GRCh38
rs778707561 Chr.3: 15667545-15667545 on Build GRCh38
rs944866914 Chr.3: 15667805-15667805 on Build GRCh38
rs746868760 Chr.3: 15667715-15667715 on Build GRCh38

View Full Product Details