Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15676710-15677227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
UniGene
Hs.335239
Gene Chromosome Location
Chr.3: 15667236-15859814 on Build GRCh38
Transcripts
NM_001195098.1 NM_001195099.1 NM_015199.3 XM_005264996.3 XM_011533540.2 XM_011533541.2 XM_011533542.2 XM_011533543.2 XM_011533545.2 XM_011533546.2 XM_011533547.2 XM_017006026.1 XM_017006027.1 XM_017006029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5059454 c.1112C>A p.T371N Chr.3: 15677046-15677046 on Build GRCh38
4770788 c.2882+1G>T p.? Chr.3: 15676973-15676973 on Build GRCh38
5863821 c.2869G>A p.A957T Chr.3: 15676987-15676987 on Build GRCh38
6600592 c.2835G>T p.K945N Chr.3: 15677021-15677021 on Build GRCh38
6996303 c.1133A>C p.E378A Chr.3: 15677025-15677025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577214765 Chr.3: 15676905-15676905 on Build GRCh38
rs370858006 Chr.3: 15677104-15677104 on Build GRCh38
rs148189292 Chr.3: 15677155-15677155 on Build GRCh38
rs540578135 Chr.3: 15676881-15676881 on Build GRCh38
rs764092703 Chr.3: 15677168-15677168 on Build GRCh38
rs1029415702 Chr.3: 15676770-15676770 on Build GRCh38
rs2470540 Chr.3: 15677145-15677145 on Build GRCh38
rs929464364 Chr.3: 15676877-15676877 on Build GRCh38
rs563607317 Chr.3: 15676988-15676988 on Build GRCh38
rs763344003 Chr.3: 15676994-15676994 on Build GRCh38
rs545958047 Chr.3: 15676954-15676954 on Build GRCh38
rs143967220 Chr.3: 15676998-15676998 on Build GRCh38
rs974029784 Chr.3: 15676847-15676847 on Build GRCh38
rs776178856 Chr.3: 15676976-15676976 on Build GRCh38
rs775161986 Chr.3: 15677036-15677036 on Build GRCh38
rs1001530026 Chr.3: 15677199-15677199 on Build GRCh38
rs959530504 Chr.3: 15676796-15676796 on Build GRCh38
rs530186897 Chr.3: 15676859-15676859 on Build GRCh38
rs942311748 Chr.3: 15676834-15676834 on Build GRCh38
rs761298767 Chr.3: 15676984-15676984 on Build GRCh38
rs991318554 Chr.3: 15676849-15676849 on Build GRCh38
rs749250771 Chr.3: 15677073-15677074 on Build GRCh38
rs973722986 Chr.3: 15677041-15677041 on Build GRCh38
rs777914019 Chr.3: 15677072-15677072 on Build GRCh38
rs1055065285 Chr.3: 15676899-15676900 on Build GRCh38
rs570353481 Chr.3: 15677183-15677183 on Build GRCh38
rs1020571264 Chr.3: 15677172-15677172 on Build GRCh38
rs767999328 Chr.3: 15676974-15676974 on Build GRCh38
rs776086026 Chr.3: 15676757-15676757 on Build GRCh38
rs755489229 Chr.3: 15677025-15677025 on Build GRCh38
rs539535013 Chr.3: 15676930-15676930 on Build GRCh38
rs374417222 Chr.3: 15677030-15677030 on Build GRCh38
rs779919230 Chr.3: 15677100-15677100 on Build GRCh38
rs375834695 Chr.3: 15677029-15677029 on Build GRCh38
rs902806954 Chr.3: 15677187-15677187 on Build GRCh38
rs149481554 Chr.3: 15676987-15676987 on Build GRCh38
rs572008889 Chr.3: 15676750-15676750 on Build GRCh38
rs756326688 Chr.3: 15676928-15676928 on Build GRCh38
rs917976043 Chr.3: 15676853-15676853 on Build GRCh38
rs552482825 Chr.3: 15677159-15677159 on Build GRCh38
rs201334606 Chr.3: 15677000-15677000 on Build GRCh38
rs1038778209 Chr.3: 15677113-15677113 on Build GRCh38
rs562323056 Chr.3: 15677062-15677062 on Build GRCh38
rs946704312 Chr.3: 15677169-15677169 on Build GRCh38
rs749512053 Chr.3: 15677079-15677079 on Build GRCh38
rs938035476 Chr.3: 15676896-15676896 on Build GRCh38
rs919438814 Chr.3: 15676869-15676869 on Build GRCh38
rs953726525 Chr.3: 15676775-15676775 on Build GRCh38
rs188908140 Chr.3: 15676891-15676891 on Build GRCh38
rs745648754 Chr.3: 15676951-15676951 on Build GRCh38
rs1046500658 Chr.3: 15676882-15676882 on Build GRCh38
rs1042342074 Chr.3: 15677205-15677205 on Build GRCh38
rs377593632 Chr.3: 15677080-15677080 on Build GRCh38
rs746472481 Chr.3: 15676860-15676860 on Build GRCh38
rs909526712 Chr.3: 15676810-15676810 on Build GRCh38
rs368294336 Chr.3: 15677039-15677039 on Build GRCh38
rs778035597 Chr.3: 15676950-15676950 on Build GRCh38
rs1010828957 Chr.3: 15676920-15676920 on Build GRCh38
rs756243817 Chr.3: 15677070-15677070 on Build GRCh38
rs998010675 Chr.3: 15676983-15676983 on Build GRCh38
rs746995363 Chr.3: 15676966-15676966 on Build GRCh38

View Full Product Details