Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15677167-15677670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
UniGene
Hs.335239
Gene Chromosome Location
Chr.3: 15667236-15859814 on Build GRCh38
Transcripts
NM_001195098.1 NM_001195099.1 NM_015199.3 XM_005264996.3 XM_011533540.2 XM_011533541.2 XM_011533542.2 XM_011533543.2 XM_011533545.2 XM_011533546.2 XM_011533547.2 XM_017006026.1 XM_017006027.1 XM_017006029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661826 c.2780T>G p.L927R Chr.3: 15677499-15677499 on Build GRCh38
5910344 c.2717-8C>T p.? Chr.3: 15677570-15677570 on Build GRCh38
4661827 c.1082T>G p.L361R Chr.3: 15677499-15677499 on Build GRCh38
5910345 c.1019-8C>T p.? Chr.3: 15677570-15677570 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032559398 Chr.3: 15677226-15677226 on Build GRCh38
rs776492974 Chr.3: 15677476-15677476 on Build GRCh38
rs182807142 Chr.3: 15677388-15677388 on Build GRCh38
rs761501087 Chr.3: 15677263-15677263 on Build GRCh38
rs186307725 Chr.3: 15677441-15677441 on Build GRCh38
rs372121402 Chr.3: 15677462-15677464 on Build GRCh38
rs141158545 Chr.3: 15677630-15677630 on Build GRCh38
rs959817147 Chr.3: 15677232-15677232 on Build GRCh38
rs772960155 Chr.3: 15677507-15677507 on Build GRCh38
rs933904612 Chr.3: 15677585-15677585 on Build GRCh38
rs34208188 Chr.3: 15677542-15677542 on Build GRCh38
rs1001530026 Chr.3: 15677199-15677199 on Build GRCh38
rs902414058 Chr.3: 15677210-15677210 on Build GRCh38
rs774497857 Chr.3: 15677240-15677240 on Build GRCh38
rs759882787 Chr.3: 15677540-15677540 on Build GRCh38
rs998041756 Chr.3: 15677227-15677227 on Build GRCh38
rs767360543 Chr.3: 15677358-15677358 on Build GRCh38
rs923936553 Chr.3: 15677544-15677544 on Build GRCh38
rs756116905 Chr.3: 15677559-15677559 on Build GRCh38
rs901637170 Chr.3: 15677640-15677640 on Build GRCh38
rs747640528 Chr.3: 15677480-15677480 on Build GRCh38
rs768636693 Chr.3: 15677436-15677436 on Build GRCh38
rs1025085125 Chr.3: 15677257-15677257 on Build GRCh38
rs972761693 Chr.3: 15677297-15677297 on Build GRCh38
rs915303376 Chr.3: 15677527-15677527 on Build GRCh38
rs754747062 Chr.3: 15677591-15677591 on Build GRCh38
rs962442858 Chr.3: 15677360-15677360 on Build GRCh38
rs116098782 Chr.3: 15677379-15677379 on Build GRCh38
rs751359425 Chr.3: 15677584-15677584 on Build GRCh38
rs564884908 Chr.3: 15677321-15677321 on Build GRCh38
rs766184935 Chr.3: 15677520-15677520 on Build GRCh38
rs548971260 Chr.3: 15677543-15677543 on Build GRCh38
rs549598148 Chr.3: 15677376-15677376 on Build GRCh38
rs67528545 Chr.3: 15677437-15677442 on Build GRCh38
rs757446419 Chr.3: 15677569-15677569 on Build GRCh38
rs985126385 Chr.3: 15677405-15677405 on Build GRCh38
rs781122085 Chr.3: 15677607-15677607 on Build GRCh38
rs368456997 Chr.3: 15677580-15677580 on Build GRCh38
rs769436794 Chr.3: 15677494-15677494 on Build GRCh38
rs1006646961 Chr.3: 15677306-15677306 on Build GRCh38
rs779074295 Chr.3: 15677570-15677570 on Build GRCh38
rs192639139 Chr.3: 15677365-15677365 on Build GRCh38
rs1017095082 Chr.3: 15677340-15677340 on Build GRCh38
rs764186168 Chr.3: 15677566-15677566 on Build GRCh38
rs1042342074 Chr.3: 15677205-15677205 on Build GRCh38
rs774748129 Chr.3: 15677521-15677521 on Build GRCh38
rs933070477 Chr.3: 15677645-15677645 on Build GRCh38
rs150736467 Chr.3: 15677460-15677462 on Build GRCh38
rs753943620 Chr.3: 15677359-15677359 on Build GRCh38
rs762717215 Chr.3: 15677513-15677513 on Build GRCh38
rs994227153 Chr.3: 15677367-15677367 on Build GRCh38

View Full Product Details