Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16264683-16265180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH3
Entrez Gene ID
285381
Gene Name
diphthamide biosynthesis 3
Gene Aliases
DELGIP, DELGIP1, DESR1, DPH3A, KTI11, ZCSL2
UniGene
Hs.388087
Gene Chromosome Location
Chr.3: 16257061-16264989 on Build GRCh38
Transcripts
NM_001047434.2 NM_206831.2 NM_138381.3 XM_017007486.1 XM_017007487.1 XM_017007488.1 XM_017007489.1 XM_017007490.1 XM_017007491.1

Gene Symbol
OXNAD1
Entrez Gene ID
92106
Gene Name
oxidoreductase NAD binding domain containing 1
Gene Aliases
-
UniGene
Hs.655449
Gene Chromosome Location
Chr.3: 16265160-16386979 on Build GRCh38
Transcripts
NM_001047434.2 NM_206831.2 NM_138381.3 XM_017007486.1 XM_017007487.1 XM_017007488.1 XM_017007489.1 XM_017007490.1 XM_017007491.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4769709 c.12T>C p.F4F Chr.3: 16264865-16264865 on Build GRCh38
1250624 c.30C>G p.I10M Chr.3: 16264847-16264847 on Build GRCh38
6269219 c.48C>A p.D16E Chr.3: 16264829-16264829 on Build GRCh38
280589 c.69C>A p.F23L Chr.3: 16264808-16264808 on Build GRCh38
6468607 c.55T>C p.S19P Chr.3: 16264822-16264822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570878564 Chr.3: 16264810-16264810 on Build GRCh38
rs1000151271 Chr.3: 16265024-16265024 on Build GRCh38
rs765917087 Chr.3: 16264737-16264737 on Build GRCh38
rs994113296 Chr.3: 16265095-16265095 on Build GRCh38
rs762737219 Chr.3: 16264814-16264814 on Build GRCh38
rs200031105 Chr.3: 16264894-16264894 on Build GRCh38
rs780226141 Chr.3: 16264791-16264791 on Build GRCh38
rs762444952 Chr.3: 16265131-16265131 on Build GRCh38
rs112129039 Chr.3: 16264953-16264953 on Build GRCh38
rs1014521098 Chr.3: 16265064-16265064 on Build GRCh38
rs745978849 Chr.3: 16264786-16264786 on Build GRCh38
rs756425218 Chr.3: 16264761-16264761 on Build GRCh38
rs1025641228 Chr.3: 16265106-16265106 on Build GRCh38
rs201354877 Chr.3: 16264806-16264806 on Build GRCh38
rs972921916 Chr.3: 16264728-16264728 on Build GRCh38
rs750975933 Chr.3: 16264862-16264862 on Build GRCh38
rs942051845 Chr.3: 16265006-16265006 on Build GRCh38
rs1056124322 Chr.3: 16265039-16265039 on Build GRCh38
rs949397237 Chr.3: 16264717-16264717 on Build GRCh38
rs979906283 Chr.3: 16264929-16264929 on Build GRCh38
rs758457515 Chr.3: 16264864-16264864 on Build GRCh38
rs767196710 Chr.3: 16264824-16264824 on Build GRCh38
rs759158161 Chr.3: 16265065-16265065 on Build GRCh38
rs781757681 Chr.3: 16264883-16264884 on Build GRCh38
rs752447007 Chr.3: 16264754-16264754 on Build GRCh38
rs577665019 Chr.3: 16265019-16265019 on Build GRCh38
rs984694988 Chr.3: 16264954-16264954 on Build GRCh38
rs932107502 Chr.3: 16264874-16264874 on Build GRCh38
rs749137653 Chr.3: 16264884-16264884 on Build GRCh38
rs201620103 Chr.3: 16264913-16264913 on Build GRCh38
rs559404922 Chr.3: 16265144-16265144 on Build GRCh38
rs769470756 Chr.3: 16264803-16264803 on Build GRCh38
rs772180775 Chr.3: 16264790-16264790 on Build GRCh38
rs563514124 Chr.3: 16265037-16265037 on Build GRCh38
rs953069946 Chr.3: 16265109-16265109 on Build GRCh38
rs186506931 Chr.3: 16264859-16264859 on Build GRCh38
rs975869067 Chr.3: 16264964-16264964 on Build GRCh38
rs764058774 Chr.3: 16265134-16265134 on Build GRCh38
rs369519531 Chr.3: 16264880-16264880 on Build GRCh38
rs1000418511 Chr.3: 16264897-16264897 on Build GRCh38
rs769679007 Chr.3: 16264719-16264719 on Build GRCh38
rs539501472 Chr.3: 16264816-16264816 on Build GRCh38
rs757721182 Chr.3: 16264856-16264856 on Build GRCh38
rs1013776989 Chr.3: 16264909-16264909 on Build GRCh38
rs776188693 Chr.3: 16264904-16264904 on Build GRCh38
rs747313966 Chr.3: 16264795-16264795 on Build GRCh38
rs762144463 Chr.3: 16264906-16264906 on Build GRCh38
rs980196569 Chr.3: 16264764-16264764 on Build GRCh38
rs751678891 Chr.3: 16264916-16264916 on Build GRCh38
rs182195034 Chr.3: 16265132-16265132 on Build GRCh38
rs984356393 Chr.3: 16265110-16265110 on Build GRCh38
rs868767787 Chr.3: 16264757-16264757 on Build GRCh38
rs1050569985 Chr.3: 16264980-16264980 on Build GRCh38
rs918828515 Chr.3: 16264947-16264947 on Build GRCh38
rs776177621 Chr.3: 16265059-16265059 on Build GRCh38
rs759211235 Chr.3: 16264749-16264749 on Build GRCh38
rs780247320 Chr.3: 16264865-16264865 on Build GRCh38
rs967254239 Chr.3: 16265102-16265102 on Build GRCh38
rs764310617 Chr.3: 16264842-16264842 on Build GRCh38
rs778269519 Chr.3: 16264762-16264762 on Build GRCh38
rs781442592 Chr.3: 16264918-16264918 on Build GRCh38
rs150059711 Chr.3: 16264779-16264779 on Build GRCh38
rs747132762 Chr.3: 16264866-16264866 on Build GRCh38
rs755237843 Chr.3: 16264871-16264871 on Build GRCh38
rs894268292 Chr.3: 16265050-16265050 on Build GRCh38
rs770886656 Chr.3: 16264885-16264885 on Build GRCh38
rs759083264 Chr.3: 16264821-16264821 on Build GRCh38
rs112968077 Chr.3: 16264978-16264978 on Build GRCh38
rs748426909 Chr.3: 16264921-16264921 on Build GRCh38
rs747431355 Chr.3: 16264970-16264970 on Build GRCh38
rs945858712 Chr.3: 16264956-16264956 on Build GRCh38
rs970676246 Chr.3: 16264943-16264943 on Build GRCh38
rs760384246 Chr.3: 16264838-16264838 on Build GRCh38
rs751125512 Chr.3: 16264745-16264745 on Build GRCh38
rs34677720 Chr.3: 16264905-16264906 on Build GRCh38
rs766296288 Chr.3: 16265016-16265016 on Build GRCh38
rs772773464 Chr.3: 16264784-16264784 on Build GRCh38
rs1050842595 Chr.3: 16264878-16264878 on Build GRCh38
rs745746351 Chr.3: 16264927-16264927 on Build GRCh38
rs755111697 Chr.3: 16264917-16264917 on Build GRCh38
rs775120003 Chr.3: 16265088-16265088 on Build GRCh38
rs773007697 Chr.3: 16264720-16264720 on Build GRCh38
rs972719470 Chr.3: 16264946-16264946 on Build GRCh38
rs766310935 Chr.3: 16264915-16264915 on Build GRCh38
rs765647025 Chr.3: 16264911-16264911 on Build GRCh38
rs374219730 Chr.3: 16264893-16264893 on Build GRCh38
rs917952166 Chr.3: 16264716-16264716 on Build GRCh38
rs373911088 Chr.3: 16264734-16264734 on Build GRCh38
rs371722277 Chr.3: 16264888-16264888 on Build GRCh38
rs931446883 Chr.3: 16264747-16264747 on Build GRCh38
rs760197436 Chr.3: 16264900-16264900 on Build GRCh38
rs775003955 Chr.3: 16264828-16264828 on Build GRCh38
rs900941333 Chr.3: 16264986-16264986 on Build GRCh38
rs972439810 Chr.3: 16264724-16264724 on Build GRCh38
rs957099848 Chr.3: 16264940-16264940 on Build GRCh38
rs1024071702 Chr.3: 16264746-16264746 on Build GRCh38
rs990884325 Chr.3: 16264928-16264928 on Build GRCh38
rs543502943 Chr.3: 16265033-16265033 on Build GRCh38
rs557705396 Chr.3: 16265011-16265011 on Build GRCh38
rs773956121 Chr.3: 16264819-16264819 on Build GRCh38
rs547736683 Chr.3: 16264770-16264770 on Build GRCh38
rs945666494 Chr.3: 16265005-16265005 on Build GRCh38
rs748543199 Chr.3: 16264924-16264925 on Build GRCh38
rs2271078 Chr.3: 16265027-16265027 on Build GRCh38
rs755323929 Chr.3: 16264884-16264884 on Build GRCh38
rs757116637 Chr.3: 16264924-16264924 on Build GRCh38
rs964221274 Chr.3: 16265127-16265127 on Build GRCh38
rs942871514 Chr.3: 16264976-16264976 on Build GRCh38
rs778858454 Chr.3: 16264925-16264925 on Build GRCh38
rs976003462 Chr.3: 16265119-16265119 on Build GRCh38
rs1019930411 Chr.3: 16265080-16265080 on Build GRCh38
rs911466541 Chr.3: 16265121-16265121 on Build GRCh38
rs111654033 Chr.3: 16265068-16265068 on Build GRCh38
rs1011214300 Chr.3: 16265077-16265077 on Build GRCh38
rs1025015456 Chr.3: 16264934-16264934 on Build GRCh38
rs1041427683 Chr.3: 16265009-16265009 on Build GRCh38
rs200723352 Chr.3: 16264765-16264765 on Build GRCh38
rs1001689030 Chr.3: 16265054-16265054 on Build GRCh38
rs754252036 Chr.3: 16264847-16264847 on Build GRCh38

View Full Product Details