The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320143.1 | NP_001307072.1 | 8 - 9 | 1220 | 82 |
NM_001320144.1 | NP_001307073.1 | 8 - 9 | 1081 | 82 | |
NM_025228.3 | NP_079504.2 | 8 - 9 | 1141 | 82 | |
NR_109871.1 | 8 - 9 | 1141 | 82 | ||
XM_011510018.2 | XP_011508320.1 | 8 - 9 | 1125 | 82 | |
XM_011510019.1 | XP_011508321.1 | 8 - 9 | 1125 | 82 | |
XM_017002399.1 | XP_016857888.1 | 8 - 9 | 1125 | 82 | |
XM_017002400.1 | XP_016857889.1 | 8 - 9 | 1125 | 82 | |
GenBank mRNA | AK122792.1 | - | 8 - 9 | 1124 | 82 |
AK125601.1 | - | 8 - 9 | 1220 | 82 | |
AK225762.1 | - | 8 - 9 | 1036 | 82 | |
AL049667.1 | - | 8 - 9 | 934 | 82 | |
BC110302.1 | - | 8 - 9 | 1107 | 82 | |
BC130417.1 | - | 7 - 8 | 821 | 82 | |
BC144139.1 | - | 7 - 8 | 821 | 82 | |
BX648450.1 | - | 8 - 9 | 1120 | 82 |