The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025258.2 | NP_079534.2 | 7 - 8 | 1327 | 62 |
XM_005249427.2 | XP_005249484.1 | 7 - 8 | 1327 | 62 | |
XM_017011327.1 | XP_016866816.1 | 6 - 7 | 1362 | 62 | |
XM_017011328.1 | XP_016866817.1 | 7 - 8 | 1473 | 62 | |
XM_017011329.1 | XP_016866818.1 | 5 - 6 | 871 | 62 | |
GenBank mRNA | AJ245418.1 | - | 2 - 3 | 197 | 62 |
AK304058.1 | - | 7 - 8 | 1187 | 62 | |
AK304266.1 | - | 6 - 7 | 1100 | 62 | |
BC114625.1 | - | 6 - 7 | 1086 | 62 |