Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
Chromosome Location
Chr.16: 74873568 - 74985146 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324171.1 NP_001311100.1 10 - 11 1014 74
NM_001324172.1 NP_001311101.1 10 - 11 1014 74
NM_030581.3 NP_085058.3 10 - 11 987 74
XM_005256146.3 XP_005256203.1 10 - 11 991 74
XM_011523332.2 XP_011521634.1 4 - 5 414 74
XM_017023667.1 XP_016879156.1 9 - 10 848 74
XM_017023668.1 XP_016879157.1 8 - 9 624 74
GenBank mRNA AB067510.1 - 5 - 6 460 74
AK302557.1 - 10 - 11 1055 74
AL562571.3 - 4 - 5 74
BC004519.2 - 10 - 11 951 74
BC014887.2 - 4 - 5 379 74

View Full Product Details