Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMGN5
Entrez Gene ID
79366
Gene Name
high mobility group nucleosome binding domain 5
Gene Aliases
NBP-45, NSBP1
Chromosome Location
Chr.X: 81113701 - 81201942 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030763.2 NP_110390.1 2 - 3 345 104
XM_006724688.3 XP_006724751.1 2 - 3 292 104
GenBank mRNA AF250329.1 - 1 - 2 84 104
AK094058.1 - 2 - 3 345 104
AK315792.1 - 1 - 2 138 104
BC005342.1 - 1 - 2 105 104
DA745749.1 - 2 - 3 314 104

View Full Product Details