The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030785.3 | NP_110412.1 | 1 - 2 | 825 | 76 |
XM_011527351.1 | XP_011525653.1 | 1 - 2 | 801 | 76 | |
GenBank mRNA | AK098837.1 | - | 1 - 2 | 770 | 76 |
AK223347.1 | - | 1 - 2 | 770 | 76 | |
AL136761.1 | - | 1 - 2 | 76 | ||
BC057785.1 | - | 1 - 2 | 653 | 76 | |
BC136351.1 | - | 1 - 2 | 706 | 76 | |
BC136352.1 | - | 1 - 2 | 706 | 76 |