The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193478.1 | NP_001180407.1 | 8 - 9 | 1541 | 77 |
NM_030911.3 | NP_112173.1 | 8 - 9 | 1541 | 77 | |
NR_036439.1 | 7 - 8 | 1363 | 77 | ||
XM_006719871.3 | XP_006719934.1 | 7 - 8 | 1310 | 77 | |
XM_011535250.2 | XP_011533552.1 | 8 - 9 | 1505 | 77 | |
XM_011535251.1 | XP_011533553.1 | 8 - 9 | 1485 | 77 | |
XM_011535253.1 | XP_011533555.1 | 8 - 9 | 1437 | 77 | |
XM_017020778.1 | XP_016876267.1 | 7 - 8 | 1244 | 77 | |
XM_017020779.1 | XP_016876268.1 | 9 - 10 | 1661 | 77 | |
XM_017020780.1 | XP_016876269.1 | 9 - 10 | 2008 | 77 | |
XM_017020781.1 | XP_016876270.1 | 8 - 9 | 1480 | 77 | |
XM_017020782.1 | XP_016876271.1 | 9 - 10 | 1584 | 77 | |
XM_017020783.1 | XP_016876272.1 | 7 - 8 | 1705 | 77 | |
XM_017020784.1 | XP_016876273.1 | 6 - 7 | 1276 | 77 | |
GenBank mRNA | AK297400.1 | - | 7 - 8 | 1327 | 77 |
AK312835.1 | - | 8 - 9 | 1511 | 77 | |
AL137554.1 | - | 2 - 3 | 217 | 77 | |
AY027525.1 | - | 8 - 9 | 1490 | 77 | |
BC048092.1 | - | 9 - 10 | 1509 | 77 | |
BC125202.1 | - | 8 - 9 | 1455 | 77 |