The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013437.1 | NP_001013455.1 | 5 - 6 | 759 | 100 |
NM_031216.3 | NP_112493.2 | 5 - 6 | 759 | 100 | |
XM_005258152.4 | XP_005258209.1 | 4 - 5 | 602 | 100 | |
XM_011525742.2 | XP_011524044.1 | 6 - 7 | 815 | 100 | |
XM_011525743.2 | XP_011524045.1 | 5 - 6 | 662 | 100 | |
XM_011525744.1 | XP_011524046.1 | 6 - 7 | 815 | 100 | |
XM_011525745.2 | XP_011524047.1 | 5 - 6 | 583 | 100 | |
XM_011525746.2 | XP_011524048.1 | 5 - 6 | 865 | 100 | |
XM_017026025.1 | XP_016881514.1 | 4 - 5 | 608 | 100 | |
XM_017026026.1 | XP_016881515.1 | 4 - 5 | 679 | 100 | |
XM_017026027.1 | XP_016881516.1 | 5 - 6 | 929 | 100 | |
GenBank mRNA | AF136976.1 | - | 5 - 6 | 727 | 100 |
AF255625.1 | - | 5 - 6 | 663 | 100 | |
AF431970.1 | - | 5 - 6 | 663 | 100 | |
AF514996.1 | - | 5 - 6 | 623 | 100 | |
AK056940.1 | - | 5 - 6 | 722 | 100 | |
AK291226.1 | - | 5 - 6 | 722 | 100 | |
AK314403.1 | - | 5 - 6 | 733 | 100 | |
BC012430.1 | - | 5 - 6 | 713 | 100 |