The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031283.2 | NP_112573.1 | 3 - 4 | 669 | 106 |
XM_006712109.2 | XP_006712172.1 | 3 - 4 | 459 | 106 | |
GenBank mRNA | AB031046.1 | - | 3 - 4 | 518 | 106 |
BC058894.1 | - | 3 - 4 | 678 | 106 |