Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- INO80B
- Entrez Gene ID
-
83444
- Gene Name
- INO80 complex subunit B
- Gene Aliases
-
HMGA1L4, HMGIYL4, IES2, PAP-1BP, PAPA-1, PAPA1, ZNHIT4, hIes2
- Chromosome Location
-
Chr.2: 74455023 - 74457960 on Build GRCh38
- Gene Symbol
- INO80B-WBP1
- Entrez Gene ID
-
100532735
- Gene Name
- INO80B-WBP1 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 74455023 - 74460891 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037849.1 |
|
4 - 5 |
635 |
61 |
View Full Product Details