Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSDMC
Entrez Gene ID
56169
Gene Name
gasdermin C
Gene Aliases
MLZE
Chromosome Location
Chr.8: 129705903 - 129786888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031415.2 NP_113603.1 10 - 11 1827 82
XM_011517161.1 XP_011515463.1 10 - 11 1567 82
XM_011517162.1 XP_011515464.1 10 - 11 1567 82
XM_011517163.2 XP_011515465.1 9 - 10 794 82
XM_011517165.1 XP_011515467.1 5 - 6 306 82
XM_011517166.2 XP_011515468.1 6 - 7 1986 82
XM_017013644.1 XP_016869133.1 10 - 11 1567 82
XM_017013645.1 XP_016869134.1 8 - 9 600 82
GenBank mRNA AB042405.1 - 10 - 11 1836 82
AJ245876.1 - 4 - 5 290 82
BC035321.1 - 10 - 11 1372 82
BC063595.1 - 10 - 11 1355 82

View Full Product Details