The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024380.1 | NP_001019551.1 | 2 - 3 | 587 | 64 |
NM_001024381.1 | NP_001019552.1 | 2 - 3 | 486 | 64 | |
NM_031940.3 | NP_114146.3 | 2 - 3 | 793 | 64 | |
NM_078473.2 | NP_510882.1 | 2 - 3 | 404 | 64 | |
XM_005273657.3 | XP_005273714.1 | 2 - 3 | 687 | 64 | |
XM_006716408.3 | XP_006716471.1 | 2 - 3 | 569 | 64 | |
XM_011544667.2 | XP_011542969.1 | 2 - 3 | 929 | 64 | |
GenBank mRNA | AF353991.1 | - | 2 - 3 | 374 | 64 |
AK075027.1 | - | 2 - 3 | 765 | 64 | |
AK075155.1 | - | 2 - 3 | 788 | 64 | |
AK225093.1 | - | 2 - 3 | 767 | 64 | |
AK314703.1 | - | 2 - 3 | 393 | 64 | |
AL834224.1 | - | 2 - 3 | 447 | 64 | |
BC004878.2 | - | 2 - 3 | 756 | 64 | |
BC109049.1 | - | 2 - 3 | 372 | 64 | |
BC109050.1 | - | 2 - 3 | 370 | 64 | |
BG424960.1 | - | - | 404 | 64 |