The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031949.4 | NP_114155.4 | 1 - 2 | 136 | 88 |
GenBank mRNA | AK093039.1 | - | 1 - 2 | 136 | 88 |
AK225984.1 | - | 1 - 2 | 136 | 88 | |
AK314450.1 | - | 1 - 2 | 113 | 88 | |
AY026506.1 | - | 1 - 2 | 108 | 88 | |
BC030650.1 | - | 1 - 2 | 126 | 88 | |
BC047411.1 | - | 1 - 2 | 114 | 88 |