The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136153.1 | NP_001129625.1 | 5 - 6 | 501 | 101 |
NM_004381.4 | NP_004372.3 | 5 - 6 | 510 | 101 | |
GenBank mRNA | BC008394.1 | - | 5 - 6 | 492 | 101 |
BC077075.1 | - | 5 - 6 | 483 | 101 | |
BT007042.1 | - | 5 - 6 | 477 | 101 | |
U31903.1 | - | 5 - 6 | 514 | 101 | |
X98054.1 | - | 5 - 6 | 510 | 101 |