The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258207.1 | NP_001245136.1 | 20 - 21 | 2025 | 75 |
NM_005428.3 | NP_005419.2 | 21 - 22 | 2121 | 75 | |
XM_005259642.1 | XP_005259699.1 | 21 - 22 | 2081 | 75 | |
GenBank mRNA | AK301128.1 | - | 20 - 21 | 1735 | 75 |
AK313307.1 | - | 21 - 22 | 2063 | 75 | |
BC013361.2 | - | 21 - 22 | 1901 | 75 | |
X16316.1 | - | 20 - 21 | 1944 | 75 | |
X83931.1 | - | 13 - 14 | 1087 | 75 |