The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199573.1 | NP_001186502.1 | 1 - 2 | 315 | 83 |
NM_006074.4 | NP_006065.2 | 1 - 2 | 315 | 83 | |
GenBank mRNA | AK290434.1 | - | 1 - 2 | 45 | 83 |
AK298715.1 | - | 1 - 2 | 101 | 83 | |
AK298934.1 | - | 1 - 2 | 316 | 83 | |
AK308221.1 | - | 1 - 2 | 135 | 83 | |
AK310098.1 | - | 1 - 2 | 135 | 83 | |
AK311659.1 | - | 1 - 2 | 136 | 83 | |
BC022281.1 | - | 1 - 2 | 122 | 83 | |
BC035582.1 | - | 1 - 2 | 88 | 83 | |
CD621768.1 | - | 1 - 2 | 216 | 83 | |
DC364428.1 | - | 1 - 2 | 292 | 83 | |
X82200.1 | - | 1 - 2 | 58 | 83 |