The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323041.1 | NP_001309970.1 | 10 - 11 | 1711 | 96 |
NM_001323042.1 | NP_001309971.1 | 10 - 11 | 1195 | 96 | |
NM_001323043.1 | NP_001309972.1 | 10 - 11 | 1596 | 96 | |
NM_001323044.1 | NP_001309973.1 | 10 - 11 | 1969 | 96 | |
NM_001323045.1 | NP_001309974.1 | 11 - 12 | 1680 | 96 | |
NM_001323046.1 | NP_001309975.1 | 7 - 8 | 1207 | 96 | |
NM_001323047.1 | NP_001309976.1 | 10 - 11 | 1711 | 96 | |
NM_001323048.1 | NP_001309977.1 | 10 - 11 | 1596 | 96 | |
NM_001323049.1 | NP_001309978.1 | 9 - 10 | 1885 | 96 | |
NM_001323050.1 | NP_001309979.1 | 11 - 12 | 1680 | 96 | |
NM_001323051.1 | NP_001309980.1 | 10 - 11 | 1969 | 96 | |
NM_001323052.1 | NP_001309981.1 | 10 - 11 | 1596 | 96 | |
NM_001323053.1 | NP_001309982.1 | 10 - 11 | 1711 | 96 | |
NM_006608.2 | NP_006599.2 | 9 - 10 | 1500 | 96 | |
NR_136564.1 | 11 - 12 | 1795 | 96 | ||
NR_136565.1 | 9 - 10 | 1436 | 96 | ||
NR_136566.1 | 10 - 11 | 1711 | 96 | ||
NR_136567.1 | 10 - 11 | 1596 | 96 | ||
NR_136568.1 | 10 - 11 | 1520 | 96 | ||
GenBank mRNA | AJ011863.1 | - | 9 - 10 | 1079 | 96 |
AK126773.1 | - | 3 - 4 | 2199 | 96 | |
AK294749.1 | - | 8 - 9 | 1066 | 96 | |
BC000842.1 | - | 3 - 4 | 315 | 96 | |
BC002447.2 | - | 10 - 11 | 1644 | 96 |