Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELF3
Entrez Gene ID
11189
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4
Chromosome Location
Chr.1: 151700058 - 151716821 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172648.3 NP_001166119.1 12 - 13 1965 59
NM_001172649.3 NP_001166120.1 11 - 12 2062 59
NM_001291106.1 NP_001278035.1 12 - 13 2209 59
NM_001291107.1 NP_001278036.1 12 - 13 2209 59
NM_007185.6 NP_009116.3 12 - 13 2212 59
XM_006711139.2 XP_006711202.1 13 - 14 1440 59
XM_006711141.2 XP_006711204.1 13 - 14 1437 59
XM_011509111.2 XP_011507413.1 11 - 12 1907 59
XM_011509112.2 XP_011507414.1 11 - 12 1303 59
XM_011509114.2 XP_011507416.1 11 - 12 1271 59
XM_017000145.1 XP_016855634.1 11 - 12 1278 59
XM_017000146.1 XP_016855635.1 11 - 12 1275 59
GenBank mRNA AB209264.1 - 12 - 13 1965 59
AF284423.1 - 3 - 4 439 59
AK298852.1 - 6 - 7 835 59
AY165003.1 - 12 - 13 2202 59
BC041088.1 - 11 - 12 1294 59
BC052491.1 - 12 - 13 2183 59
BC068008.1 - 11 - 12 1239 59
BC104758.1 - 12 - 13 1475 59
BC143226.1 - 12 - 13 1472 59
U80746.1 - 9 - 10 1091 59
U80759.1 - 9 - 10 1098 59

View Full Product Details