The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017521.2 | NP_059991.1 | 2 - 3 | 710 | 129 |
GenBank mRNA | BC023511.2 | - | 2 - 3 | 605 | 129 |
Y08976.1 | - | 2 - 3 | 712 | 129 |