Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109506741-109507238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO1
Entrez Gene ID
57463
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
ALI2, AMIGO, AMIGO-1
UniGene
Hs.726479
Gene Chromosome Location
Chr.1: 109504178-109509742 on Build GRCh38
Transcripts
NM_020703.2 XM_011541812.2

SNP Detail

SNP ID Chromosome Location
rs900698690 Chr.1: 109507155-109507155 on Build GRCh38
rs1039140227 Chr.1: 109507142-109507142 on Build GRCh38
rs146565168 Chr.1: 109507013-109507013 on Build GRCh38
rs1054794778 Chr.1: 109507038-109507038 on Build GRCh38
rs555829204 Chr.1: 109507046-109507046 on Build GRCh38
rs41301283 Chr.1: 109506835-109506835 on Build GRCh38
rs893498599 Chr.1: 109507041-109507041 on Build GRCh38
rs886087555 Chr.1: 109507179-109507179 on Build GRCh38
rs950237831 Chr.1: 109506983-109506983 on Build GRCh38
rs890744697 Chr.1: 109507128-109507128 on Build GRCh38
rs908121461 Chr.1: 109506969-109506969 on Build GRCh38
rs944969985 Chr.1: 109507131-109507132 on Build GRCh38
rs984074550 Chr.1: 109506968-109506968 on Build GRCh38
rs946413316 Chr.1: 109507048-109507048 on Build GRCh38
rs1009061885 Chr.1: 109506769-109506769 on Build GRCh38
rs1008737407 Chr.1: 109506843-109506843 on Build GRCh38
rs1020780240 Chr.1: 109506795-109506795 on Build GRCh38
rs937765907 Chr.1: 109507037-109507037 on Build GRCh38
rs915215886 Chr.1: 109507097-109507097 on Build GRCh38
rs116785924 Chr.1: 109507212-109507212 on Build GRCh38
rs974703350 Chr.1: 109506888-109506888 on Build GRCh38
rs1018764059 Chr.1: 109507165-109507165 on Build GRCh38
rs1018962036 Chr.1: 109506880-109506880 on Build GRCh38
rs996249523 Chr.1: 109507186-109507186 on Build GRCh38
rs867744058 Chr.1: 109507057-109507057 on Build GRCh38
rs981618374 Chr.1: 109507011-109507011 on Build GRCh38
rs945396519 Chr.1: 109507103-109507103 on Build GRCh38
rs555386643 Chr.1: 109507106-109507106 on Build GRCh38
rs142866212 Chr.1: 109506882-109506882 on Build GRCh38
rs535697178 Chr.1: 109507056-109507056 on Build GRCh38
rs141240479 Chr.1: 109507181-109507181 on Build GRCh38
rs952320754 Chr.1: 109506945-109506945 on Build GRCh38
rs1027636758 Chr.1: 109506920-109506920 on Build GRCh38
rs950449110 Chr.1: 109506971-109506971 on Build GRCh38
rs185803916 Chr.1: 109506885-109506885 on Build GRCh38
rs983489370 Chr.1: 109507213-109507213 on Build GRCh38
rs540599876 Chr.1: 109507211-109507211 on Build GRCh38
rs1027847139 Chr.1: 109507178-109507178 on Build GRCh38

View Full Product Details