Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109623898-109624389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD2
Entrez Gene ID
271
Gene Name
adenosine monophosphate deaminase 2
Gene Aliases
PCH9, SPG63
UniGene
Hs.82927
Gene Chromosome Location
Chr.1: 109619813-109632055 on Build GRCh38
Transcripts
NM_001257360.1 NM_001257361.1 NM_001308170.1 NM_004037.7 NM_139156.3

SNP Detail

SNP ID Chromosome Location
rs933423372 Chr.1: 109624328-109624328 on Build GRCh38
rs908469359 Chr.1: 109624329-109624331 on Build GRCh38
rs980086680 Chr.1: 109623946-109623946 on Build GRCh38
rs990458449 Chr.1: 109624017-109624017 on Build GRCh38
rs763841031 Chr.1: 109624299-109624299 on Build GRCh38
rs925225412 Chr.1: 109624302-109624302 on Build GRCh38
rs935240312 Chr.1: 109624307-109624307 on Build GRCh38
rs1029002636 Chr.1: 109624110-109624110 on Build GRCh38
rs992286155 Chr.1: 109624207-109624207 on Build GRCh38
rs551941798 Chr.1: 109624171-109624171 on Build GRCh38
rs868152382 Chr.1: 109624155-109624155 on Build GRCh38
rs1034394577 Chr.1: 109624149-109624149 on Build GRCh38
rs918373390 Chr.1: 109623934-109623934 on Build GRCh38
rs898181600 Chr.1: 109624105-109624105 on Build GRCh38
rs377400912 Chr.1: 109623969-109623969 on Build GRCh38
rs375266249 Chr.1: 109624117-109624117 on Build GRCh38
rs559833451 Chr.1: 109624122-109624122 on Build GRCh38
rs367898051 Chr.1: 109624109-109624109 on Build GRCh38
rs1046187627 Chr.1: 109624125-109624125 on Build GRCh38
rs543669579 Chr.1: 109623964-109623964 on Build GRCh38
rs960521529 Chr.1: 109624177-109624177 on Build GRCh38
rs374024570 Chr.1: 109624223-109624223 on Build GRCh38
rs886989864 Chr.1: 109624111-109624111 on Build GRCh38
rs756953746 Chr.1: 109624367-109624367 on Build GRCh38
rs947982814 Chr.1: 109624012-109624012 on Build GRCh38
rs971431276 Chr.1: 109623941-109623941 on Build GRCh38
rs1016495237 Chr.1: 109624129-109624129 on Build GRCh38
rs369969709 Chr.1: 109624069-109624069 on Build GRCh38
rs1056172348 Chr.1: 109623950-109623950 on Build GRCh38
rs542551412 Chr.1: 109624271-109624271 on Build GRCh38
rs916241636 Chr.1: 109623966-109623966 on Build GRCh38
rs957583241 Chr.1: 109624010-109624010 on Build GRCh38
rs889133117 Chr.1: 109624366-109624366 on Build GRCh38
rs370440300 Chr.1: 109624039-109624039 on Build GRCh38
rs962277204 Chr.1: 109624132-109624132 on Build GRCh38
rs1050476381 Chr.1: 109624358-109624358 on Build GRCh38
rs866040468 Chr.1: 109624143-109624143 on Build GRCh38
rs972283678 Chr.1: 109624144-109624144 on Build GRCh38
rs753590349 Chr.1: 109624359-109624359 on Build GRCh38
rs1036265832 Chr.1: 109624148-109624148 on Build GRCh38
rs939080359 Chr.1: 109623949-109623949 on Build GRCh38
rs563979320 Chr.1: 109624016-109624016 on Build GRCh38
rs986929352 Chr.1: 109624082-109624082 on Build GRCh38
rs912761751 Chr.1: 109624281-109624281 on Build GRCh38
rs942396280 Chr.1: 109624098-109624098 on Build GRCh38
rs964186664 Chr.1: 109624297-109624297 on Build GRCh38
rs901638544 Chr.1: 109624316-109624316 on Build GRCh38
rs1052308427 Chr.1: 109624313-109624313 on Build GRCh38
rs901963579 Chr.1: 109624101-109624101 on Build GRCh38
rs577850603 Chr.1: 109623937-109623937 on Build GRCh38
rs766732695 Chr.1: 109624042-109624042 on Build GRCh38
rs947702500 Chr.1: 109624248-109624248 on Build GRCh38
rs142058911 Chr.1: 109624157-109624157 on Build GRCh38
rs912331345 Chr.1: 109624091-109624091 on Build GRCh38
rs1006496113 Chr.1: 109624126-109624126 on Build GRCh38
rs181281515 Chr.1: 109624243-109624243 on Build GRCh38

View Full Product Details