The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014465.3 | NP_055280.2 | 4 - 5 | 672 | 106 |
XM_005265676.3 | XP_005265733.1 | 5 - 6 | 587 | 106 | |
XM_005265677.3 | XP_005265734.1 | 5 - 6 | 787 | 106 | |
XM_005265678.3 | XP_005265735.1 | 4 - 5 | 540 | 106 | |
XM_011531873.2 | XP_011530175.1 | 3 - 4 | 411 | 106 | |
GenBank mRNA | BC010895.1 | - | 4 - 5 | 674 | 106 |
D89479.1 | - | 3 - 4 | 398 | 106 | |
U95726.1 | - | 3 - 4 | 389 | 106 |