Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD34
Entrez Gene ID
947
Gene Name
CD34 molecule
Gene Aliases
-
Chromosome Location
Chr.1: 207886538 - 207911338 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025109.1 NP_001020280.1 3 - 4 778 90
NM_001773.2 NP_001764.1 3 - 4 778 90
GenBank mRNA AF523361.1 - 3 - 4 591 90
AF538601.1 - 2 - 3 429 90
AK291529.1 - 3 - 4 531 90
AK293030.1 - 3 - 4 555 90
AK300835.1 - 4 - 5 595 90
BC039146.1 - 3 - 4 528 90
M81104.1 - 3 - 4 777 90
S53910.1 - 3 - 4 610 90
S53911.1 - 3 - 4 610 90

View Full Product Details