Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SULT1A2
- Entrez Gene ID
-
6799
- Gene Name
- sulfotransferase family 1A member 2
- Gene Aliases
-
HAST4, P-PST, ST1A2, STP2, TSPST2
- Chromosome Location
-
Chr.16: 28591943 - 28597079 on Build GRCh38
- Gene Symbol
- SLX1A-SULT1A3
- Entrez Gene ID
-
100526830
- Gene Name
- SLX1A-SULT1A3 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 30194433 - 30204329 on Build GRCh38
- Gene Symbol
- SLX1B-SULT1A4
- Entrez Gene ID
-
100526831
- Gene Name
- SLX1B-SULT1A4 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 29455091 - 29464980 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037609.1 |
|
9 - 10 |
1517 |
72 |
- Gene Symbol
- SULT1A1
- Entrez Gene ID
-
6817
- Gene Name
- sulfotransferase family 1A member 1
- Gene Aliases
-
HAST1/HAST2, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1
- Chromosome Location
-
Chr.16: 28605587 - 28623586 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB062428.1 |
- |
5 - 6 |
652 |
72 |
L10819.1 |
- |
6 - 7 |
634 |
72 |
L19955.1 |
- |
5 - 6 |
637 |
72 |
U09031.1 |
- |
5 - 6 |
637 |
72 |
X84654.1 |
- |
5 - 6 |
610 |
72 |
- Gene Symbol
- SULT1A3
- Entrez Gene ID
-
6818
- Gene Name
- sulfotransferase family 1A member 3
- Gene Aliases
-
HAST, HAST3, M-PST, ST1A3, ST1A3/ST1A4, ST1A5, STM, TL-PST
- Chromosome Location
-
Chr.16: 30199228 - 30204329 on Build GRCh38
- Gene Symbol
- SULT1A4
- Entrez Gene ID
-
445329
- Gene Name
- sulfotransferase family 1A member 4
- Gene Aliases
-
HAST3, M-PST, ST1A3/ST1A4, ST1A4, TL-PST
- Chromosome Location
-
Chr.16: 29459886 - 29464980 on Build GRCh38
View Full Product Details