Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113897384-113897885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1C
Entrez Gene ID
54762
Gene Name
GRAM domain containing 1C
Gene Aliases
-
UniGene
Hs.24583
Gene Chromosome Location
Chr.3: 113828182-113947174 on Build GRCh38
Transcripts
NM_001172105.1 NM_017577.4 XM_005247546.2 XM_005247547.1 XM_011512930.1 XM_011512931.1 XM_017006646.1 XM_017006647.1 XM_017006648.1 XM_017006649.1

SNP Detail

SNP ID Chromosome Location
rs747342059 Chr.3: 113897675-113897675 on Build GRCh38
rs908183913 Chr.3: 113897672-113897672 on Build GRCh38
rs937208261 Chr.3: 113897435-113897435 on Build GRCh38
rs1024599440 Chr.3: 113897616-113897616 on Build GRCh38
rs545509022 Chr.3: 113897746-113897746 on Build GRCh38
rs932274960 Chr.3: 113897784-113897784 on Build GRCh38
rs1055994847 Chr.3: 113897438-113897438 on Build GRCh38
rs531308295 Chr.3: 113897513-113897513 on Build GRCh38
rs750355138 Chr.3: 113897453-113897453 on Build GRCh38
rs772473174 Chr.3: 113897440-113897442 on Build GRCh38
rs903206575 Chr.3: 113897500-113897500 on Build GRCh38
rs895573481 Chr.3: 113897444-113897444 on Build GRCh38
rs114227162 Chr.3: 113897493-113897493 on Build GRCh38
rs1018893831 Chr.3: 113897688-113897688 on Build GRCh38
rs143734581 Chr.3: 113897737-113897737 on Build GRCh38
rs1032902642 Chr.3: 113897543-113897543 on Build GRCh38
rs958620876 Chr.3: 113897587-113897587 on Build GRCh38
rs973717767 Chr.3: 113897683-113897683 on Build GRCh38
rs976829156 Chr.3: 113897825-113897825 on Build GRCh38
rs962324372 Chr.3: 113897677-113897677 on Build GRCh38
rs999836191 Chr.3: 113897514-113897514 on Build GRCh38
rs958494787 Chr.3: 113897589-113897589 on Build GRCh38
rs1032616506 Chr.3: 113897562-113897562 on Build GRCh38
rs971164686 Chr.3: 113897634-113897634 on Build GRCh38
rs912074101 Chr.3: 113897828-113897828 on Build GRCh38
rs374255832 Chr.3: 113897800-113897801 on Build GRCh38
rs1012715358 Chr.3: 113897599-113897600 on Build GRCh38
rs920805537 Chr.3: 113897772-113897772 on Build GRCh38
rs1007068156 Chr.3: 113897651-113897651 on Build GRCh38
rs1020041798 Chr.3: 113897485-113897485 on Build GRCh38
rs767397749 Chr.3: 113897452-113897452 on Build GRCh38
rs574295772 Chr.3: 113897479-113897479 on Build GRCh38
rs541494688 Chr.3: 113897797-113897798 on Build GRCh38
rs1009429852 Chr.3: 113897460-113897460 on Build GRCh38
rs559551647 Chr.3: 113897669-113897669 on Build GRCh38
rs563699252 Chr.3: 113897834-113897834 on Build GRCh38
rs986306157 Chr.3: 113897803-113897803 on Build GRCh38

View Full Product Details