Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129304509-129305017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMCES
Entrez Gene ID
56941
Gene Name
5-hydroxymethylcytosine (hmC) binding, ES cell-specific
Gene Aliases
C3orf37, DC12, SRAPD1
UniGene
Hs.458320
Gene Chromosome Location
Chr.3: 129278806-129305293 on Build GRCh38
Transcripts
NM_001006109.1 NM_020187.2 XM_005247636.3 XM_005247637.3 XM_017006877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
419845 c.922G>C p.E308Q Chr.3: 129304682-129304682 on Build GRCh38
5455132 c.984A>C p.R328S Chr.3: 129304744-129304744 on Build GRCh38
419844 c.1014G>A p.L338L Chr.3: 129304774-129304774 on Build GRCh38
3060046 c.910C>T p.P304S Chr.3: 129304670-129304670 on Build GRCh38
7082106 c.855G>C p.Q285H Chr.3: 129304615-129304615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755963170 Chr.3: 129304814-129304815 on Build GRCh38
rs755882126 Chr.3: 129304704-129304704 on Build GRCh38
rs781009402 Chr.3: 129304566-129304566 on Build GRCh38
rs1049719254 Chr.3: 129304716-129304716 on Build GRCh38
rs779777441 Chr.3: 129304707-129304707 on Build GRCh38
rs1021879751 Chr.3: 129304914-129304914 on Build GRCh38
rs376431637 Chr.3: 129304807-129304807 on Build GRCh38
rs976388591 Chr.3: 129304535-129304535 on Build GRCh38
rs757155780 Chr.3: 129304562-129304562 on Build GRCh38
rs147986626 Chr.3: 129304648-129304648 on Build GRCh38
rs140047162 Chr.3: 129304813-129304813 on Build GRCh38
rs186347235 Chr.3: 129304944-129304944 on Build GRCh38
rs754787811 Chr.3: 129304826-129304826 on Build GRCh38
rs961147566 Chr.3: 129304897-129304897 on Build GRCh38
rs757758034 Chr.3: 129304650-129304650 on Build GRCh38
rs1015358157 Chr.3: 129304796-129304796 on Build GRCh38
rs571624918 Chr.3: 129304649-129304649 on Build GRCh38
rs751603398 Chr.3: 129304670-129304670 on Build GRCh38
rs141661279 Chr.3: 129304647-129304647 on Build GRCh38
rs757343424 Chr.3: 129304682-129304682 on Build GRCh38
rs771547594 Chr.3: 129304763-129304763 on Build GRCh38
rs925825076 Chr.3: 129304979-129304979 on Build GRCh38
rs111488794 Chr.3: 129304733-129304733 on Build GRCh38
rs555310835 Chr.3: 129304917-129304917 on Build GRCh38
rs764250956 Chr.3: 129304658-129304658 on Build GRCh38
rs60527165 Chr.3: 129304787-129304787 on Build GRCh38
rs990447353 Chr.3: 129304854-129304854 on Build GRCh38
rs775094891 Chr.3: 129304585-129304585 on Build GRCh38
rs373030578 Chr.3: 129304856-129304856 on Build GRCh38
rs777060415 Chr.3: 129304627-129304627 on Build GRCh38
rs945449850 Chr.3: 129304676-129304676 on Build GRCh38
rs59474207 Chr.3: 129304699-129304699 on Build GRCh38
rs997919171 Chr.3: 129304905-129304905 on Build GRCh38
rs768287038 Chr.3: 129304715-129304715 on Build GRCh38
rs537995688 Chr.3: 129304554-129304554 on Build GRCh38
rs766851499 Chr.3: 129304623-129304623 on Build GRCh38
rs143096984 Chr.3: 129304785-129304785 on Build GRCh38
rs369963919 Chr.3: 129304808-129304808 on Build GRCh38
rs777675756 Chr.3: 129304847-129304847 on Build GRCh38
rs747077575 Chr.3: 129304557-129304557 on Build GRCh38
rs761981342 Chr.3: 129304878-129304878 on Build GRCh38
rs749113566 Chr.3: 129304710-129304710 on Build GRCh38
rs888546594 Chr.3: 129304730-129304730 on Build GRCh38
rs760166437 Chr.3: 129304778-129304778 on Build GRCh38
rs768320589 Chr.3: 129304617-129304617 on Build GRCh38
rs558051897 Chr.3: 129304784-129304784 on Build GRCh38
rs769646873 Chr.3: 129304575-129304575 on Build GRCh38
rs747794464 Chr.3: 129304875-129304875 on Build GRCh38
rs746198617 Chr.3: 129304746-129304746 on Build GRCh38
rs753570955 Chr.3: 129304817-129304817 on Build GRCh38
rs745652637 Chr.3: 129304568-129304568 on Build GRCh38
rs36104687 Chr.3: 129304778-129304779 on Build GRCh38
rs372605139 Chr.3: 129304757-129304757 on Build GRCh38
rs759973871 Chr.3: 129304630-129304630 on Build GRCh38
rs764367352 Chr.3: 129304788-129304788 on Build GRCh38
rs756006412 Chr.3: 129304809-129304809 on Build GRCh38
rs773873060 Chr.3: 129304621-129304621 on Build GRCh38
rs777161597 Chr.3: 129304769-129304769 on Build GRCh38
rs761233096 Chr.3: 129304622-129304622 on Build GRCh38
rs779376468 Chr.3: 129304666-129304666 on Build GRCh38
rs748991945 Chr.3: 129304600-129304600 on Build GRCh38
rs751798055 Chr.3: 129304799-129304799 on Build GRCh38
rs978240979 Chr.3: 129304974-129304974 on Build GRCh38
rs13255 Chr.3: 129304962-129304962 on Build GRCh38
rs773888943 Chr.3: 129304750-129304750 on Build GRCh38
rs553051089 Chr.3: 129304962-129304963 on Build GRCh38

View Full Product Details