Product Details

Assay Reference Genome
Location

Chr.13:50929586 on build GRCh38
Cytoband
13q14.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
79621
Gene Symbol
RNASEH2B
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
Location
Chr.13:50909678-50970462 on build GRCh38
Assay Gene Location
Overlaps Exon 4 - Intron 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
RNASEH2B NM_001142279.2 NP_001135751.1
NM_024570.3 NP_078846.2
XM_006719867.3 XP_006719930.1
XM_011535229.1 XP_011533531.1
XM_011535230.2 XP_011533532.1
XM_011535231.2 XP_011533533.1
XM_011535233.2 XP_011533535.1
XM_017020747.1 XP_016876236.1
AK021774.1 BAB13892.1
AK223340.1 BAD97060.1
AY764036.1 AAX13343.1
BC001397.2 AAH01397.1
BC005088.1
BC007332.2 AAH07332.1
BC010174.2 AAH10174.1
BC036744.1 AAH36744.1
BX382357.2
DB221997.1
DT216141.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv561618 Chr13:49907577 - 51521449 on Build GRCh38 Loss DLEU7 ST13P4 LINC00371 DLEU1-AS1 KCNRG DLEU2 RNASEH2B-AS1 MIR16-1 SERPINE3 INTS6 DLEU1 RNASEH2B MIR3613 FAM124A TRIM13 MIR5693 MIR15A SPRYD7 DLEU7-AS1 GUCY1B2 INTS6-AS1

View Full Product Details