Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133400206-133400705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BFSP2
Entrez Gene ID
8419
Gene Name
beaded filament structural protein 2
Gene Aliases
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN
UniGene
Hs.659862
Gene Chromosome Location
Chr.3: 133397882-133475212 on Build GRCh38
Transcripts
NM_003571.3 XM_017007315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611405 c.182C>T p.A61V Chr.3: 133400265-133400265 on Build GRCh38
5445372 c.489+6T>C p.? Chr.3: 133400578-133400578 on Build GRCh38
1670580 c.397G>A p.E133K Chr.3: 133400480-133400480 on Build GRCh38
3914959 c.234C>T p.L78L Chr.3: 133400317-133400317 on Build GRCh38
3974171 c.409C>T p.R137W Chr.3: 133400492-133400492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200369728 Chr.3: 133400348-133400348 on Build GRCh38
rs758203603 Chr.3: 133400267-133400267 on Build GRCh38
rs749995797 Chr.3: 133400419-133400419 on Build GRCh38
rs758115667 Chr.3: 133400339-133400339 on Build GRCh38
rs555303536 Chr.3: 133400668-133400668 on Build GRCh38
rs748115575 Chr.3: 133400368-133400368 on Build GRCh38
rs759174901 Chr.3: 133400528-133400528 on Build GRCh38
rs766843026 Chr.3: 133400306-133400306 on Build GRCh38
rs768109082 Chr.3: 133400313-133400313 on Build GRCh38
rs375213601 Chr.3: 133400580-133400580 on Build GRCh38
rs34163197 Chr.3: 133400452-133400452 on Build GRCh38
rs567594585 Chr.3: 133400648-133400648 on Build GRCh38
rs745614005 Chr.3: 133400266-133400266 on Build GRCh38
rs752477155 Chr.3: 133400543-133400543 on Build GRCh38
rs774824709 Chr.3: 133400391-133400391 on Build GRCh38
rs375248662 Chr.3: 133400268-133400268 on Build GRCh38
rs145934653 Chr.3: 133400329-133400329 on Build GRCh38
rs756062657 Chr.3: 133400486-133400486 on Build GRCh38
rs886058009 Chr.3: 133400505-133400505 on Build GRCh38
rs957602637 Chr.3: 133400345-133400345 on Build GRCh38
rs769854464 Chr.3: 133400292-133400292 on Build GRCh38
rs769098210 Chr.3: 133400604-133400604 on Build GRCh38
rs775787257 Chr.3: 133400503-133400503 on Build GRCh38
rs763430287 Chr.3: 133400231-133400231 on Build GRCh38
rs991256139 Chr.3: 133400429-133400429 on Build GRCh38
rs770418692 Chr.3: 133400616-133400616 on Build GRCh38
rs766932682 Chr.3: 133400232-133400232 on Build GRCh38
rs773954258 Chr.3: 133400282-133400282 on Build GRCh38
rs771326028 Chr.3: 133400376-133400376 on Build GRCh38
rs780433315 Chr.3: 133400597-133400597 on Build GRCh38
rs540517188 Chr.3: 133400351-133400351 on Build GRCh38
rs1052451744 Chr.3: 133400459-133400459 on Build GRCh38
rs779218350 Chr.3: 133400579-133400579 on Build GRCh38
rs763339964 Chr.3: 133400304-133400304 on Build GRCh38
rs190619931 Chr.3: 133400491-133400491 on Build GRCh38
rs772409951 Chr.3: 133400408-133400408 on Build GRCh38
rs774912497 Chr.3: 133400309-133400309 on Build GRCh38
rs144399466 Chr.3: 133400519-133400519 on Build GRCh38
rs536551983 Chr.3: 133400650-133400650 on Build GRCh38
rs761163518 Chr.3: 133400416-133400416 on Build GRCh38
rs1039029166 Chr.3: 133400247-133400247 on Build GRCh38
rs773803150 Chr.3: 133400621-133400621 on Build GRCh38
rs145301682 Chr.3: 133400238-133400238 on Build GRCh38
rs185816798 Chr.3: 133400462-133400462 on Build GRCh38
rs377423839 Chr.3: 133400418-133400418 on Build GRCh38
rs117842116 Chr.3: 133400513-133400513 on Build GRCh38
rs764739383 Chr.3: 133400327-133400327 on Build GRCh38
rs778659632 Chr.3: 133400249-133400249 on Build GRCh38
rs749505438 Chr.3: 133400374-133400374 on Build GRCh38
rs779814177 Chr.3: 133400342-133400342 on Build GRCh38
rs762574725 Chr.3: 133400432-133400432 on Build GRCh38
rs776079190 Chr.3: 133400412-133400412 on Build GRCh38
rs748742742 Chr.3: 133400615-133400615 on Build GRCh38
rs148759360 Chr.3: 133400520-133400520 on Build GRCh38
rs752114772 Chr.3: 133400237-133400237 on Build GRCh38
rs781206833 Chr.3: 133400365-133400365 on Build GRCh38
rs142499840 Chr.3: 133400310-133400310 on Build GRCh38
rs142404860 Chr.3: 133400541-133400541 on Build GRCh38
rs746947914 Chr.3: 133400270-133400270 on Build GRCh38
rs754844945 Chr.3: 133400350-133400350 on Build GRCh38
rs756829125 Chr.3: 133400246-133400246 on Build GRCh38
rs1020307483 Chr.3: 133400614-133400614 on Build GRCh38
rs776554938 Chr.3: 133400280-133400280 on Build GRCh38
rs749731530 Chr.3: 133400572-133400572 on Build GRCh38
rs777149830 Chr.3: 133400609-133400609 on Build GRCh38
rs112142146 Chr.3: 133400245-133400245 on Build GRCh38
rs756099408 Chr.3: 133400544-133400544 on Build GRCh38
rs765954213 Chr.3: 133400523-133400523 on Build GRCh38
rs748304663 Chr.3: 133400283-133400283 on Build GRCh38
rs139476224 Chr.3: 133400449-133400449 on Build GRCh38
rs889324481 Chr.3: 133400505-133400505 on Build GRCh38
rs80176276 Chr.3: 133400493-133400493 on Build GRCh38
rs1020694617 Chr.3: 133400349-133400349 on Build GRCh38
rs746162167 Chr.3: 133400494-133400494 on Build GRCh38
rs761232858 Chr.3: 133400517-133400517 on Build GRCh38
rs1051275327 Chr.3: 133400507-133400507 on Build GRCh38
rs369202812 Chr.3: 133400305-133400305 on Build GRCh38
rs758613212 Chr.3: 133400583-133400583 on Build GRCh38
rs751224902 Chr.3: 133400524-133400524 on Build GRCh38
rs886058008 Chr.3: 133400334-133400335 on Build GRCh38
rs773577959 Chr.3: 133400298-133400298 on Build GRCh38
rs778217297 Chr.3: 133400308-133400322 on Build GRCh38
rs756814292 Chr.3: 133400318-133400318 on Build GRCh38
rs769918976 Chr.3: 133400369-133400369 on Build GRCh38
rs768699867 Chr.3: 133400279-133400279 on Build GRCh38
rs780921530 Chr.3: 133400454-133400454 on Build GRCh38
rs749419128 Chr.3: 133400492-133400492 on Build GRCh38
rs759975775 Chr.3: 133400394-133400394 on Build GRCh38
rs909038266 Chr.3: 133400673-133400673 on Build GRCh38
rs547603725 Chr.3: 133400633-133400633 on Build GRCh38
rs915826086 Chr.3: 133400631-133400631 on Build GRCh38
rs753929115 Chr.3: 133400572-133400572 on Build GRCh38
rs772491818 Chr.3: 133400496-133400496 on Build GRCh38
rs565129712 Chr.3: 133400373-133400373 on Build GRCh38
rs891311065 Chr.3: 133400515-133400515 on Build GRCh38
rs754811534 Chr.3: 133400453-133400453 on Build GRCh38
rs747379751 Chr.3: 133400598-133400598 on Build GRCh38
rs371179115 Chr.3: 133400425-133400425 on Build GRCh38
rs757416644 Chr.3: 133400575-133400575 on Build GRCh38
rs373365813 Chr.3: 133400479-133400479 on Build GRCh38

View Full Product Details