Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133448400-133448907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BFSP2
Entrez Gene ID
8419
Gene Name
beaded filament structural protein 2
Gene Aliases
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN
UniGene
Hs.659862
Gene Chromosome Location
Chr.3: 133397882-133475212 on Build GRCh38
Transcripts
NM_003571.3 XM_017007315.1 NR_135276.1 NR_135277.1 NR_135278.1

Gene Symbol
BFSP2-AS1
Entrez Gene ID
85003
Gene Name
BFSP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.659202
Gene Chromosome Location
Chr.3: 133426402-133491146 on Build GRCh38
Transcripts
NM_003571.3 XM_017007315.1 NR_135276.1 NR_135277.1 NR_135278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1670581 c.585G>T p.E195D Chr.3: 133448501-133448501 on Build GRCh38
3927353 c.651T>C p.N217N Chr.3: 133448567-133448567 on Build GRCh38
6611408 c.632A>G p.K211R Chr.3: 133448548-133448548 on Build GRCh38
6428113 c.654G>T p.L218F Chr.3: 133448570-133448570 on Build GRCh38
4455180 c.693A>G p.K231K Chr.3: 133448609-133448609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199755934 Chr.3: 133448583-133448583 on Build GRCh38
rs774368053 Chr.3: 133448651-133448651 on Build GRCh38
rs973164143 Chr.3: 133448690-133448690 on Build GRCh38
rs557265408 Chr.3: 133448560-133448560 on Build GRCh38
rs1043631012 Chr.3: 133448750-133448750 on Build GRCh38
rs763009050 Chr.3: 133448537-133448537 on Build GRCh38
rs745318662 Chr.3: 133448442-133448442 on Build GRCh38
rs748196534 Chr.3: 133448725-133448725 on Build GRCh38
rs747906000 Chr.3: 133448628-133448628 on Build GRCh38
rs121908938 Chr.3: 133448613-133448615 on Build GRCh38
rs749153171 Chr.3: 133448639-133448639 on Build GRCh38
rs754287032 Chr.3: 133448599-133448599 on Build GRCh38
rs901605072 Chr.3: 133448553-133448553 on Build GRCh38
rs916405063 Chr.3: 133448740-133448740 on Build GRCh38
rs2699873 Chr.3: 133448830-133448830 on Build GRCh38
rs990902025 Chr.3: 133448734-133448734 on Build GRCh38
rs771496555 Chr.3: 133448461-133448461 on Build GRCh38
rs369082319 Chr.3: 133448465-133448465 on Build GRCh38
rs537669301 Chr.3: 133448823-133448823 on Build GRCh38
rs928044226 Chr.3: 133448719-133448719 on Build GRCh38
rs779677903 Chr.3: 133448603-133448603 on Build GRCh38
rs886058011 Chr.3: 133448617-133448617 on Build GRCh38
rs774146557 Chr.3: 133448517-133448517 on Build GRCh38
rs1035516739 Chr.3: 133448877-133448877 on Build GRCh38
rs147660299 Chr.3: 133448554-133448554 on Build GRCh38
rs1002734833 Chr.3: 133448875-133448875 on Build GRCh38
rs868218045 Chr.3: 133448622-133448622 on Build GRCh38
rs566476785 Chr.3: 133448600-133448600 on Build GRCh38
rs753021768 Chr.3: 133448563-133448563 on Build GRCh38
rs765038524 Chr.3: 133448514-133448518 on Build GRCh38
rs981205207 Chr.3: 133448429-133448429 on Build GRCh38
rs771781592 Chr.3: 133448505-133448505 on Build GRCh38
rs984382324 Chr.3: 133448633-133448633 on Build GRCh38
rs767661946 Chr.3: 133448665-133448665 on Build GRCh38
rs918764832 Chr.3: 133448647-133448647 on Build GRCh38
rs961386266 Chr.3: 133448648-133448648 on Build GRCh38
rs761016052 Chr.3: 133448696-133448696 on Build GRCh38
rs1040451853 Chr.3: 133448781-133448781 on Build GRCh38
rs766368338 Chr.3: 133448540-133448540 on Build GRCh38
rs775704482 Chr.3: 133448666-133448666 on Build GRCh38
rs769485449 Chr.3: 133448631-133448631 on Build GRCh38
rs749155249 Chr.3: 133448489-133448489 on Build GRCh38
rs868716956 Chr.3: 133448514-133448515 on Build GRCh38
rs189376399 Chr.3: 133448512-133448512 on Build GRCh38
rs886058012 Chr.3: 133448638-133448638 on Build GRCh38
rs2859678 Chr.3: 133448652-133448652 on Build GRCh38
rs2276737 Chr.3: 133448519-133448519 on Build GRCh38
rs889945980 Chr.3: 133448858-133448858 on Build GRCh38
rs761633560 Chr.3: 133448685-133448687 on Build GRCh38
rs775149826 Chr.3: 133448506-133448507 on Build GRCh38
rs1050206358 Chr.3: 133448857-133448857 on Build GRCh38
rs923873282 Chr.3: 133448755-133448755 on Build GRCh38
rs995924992 Chr.3: 133448821-133448821 on Build GRCh38
rs779767821 Chr.3: 133448464-133448464 on Build GRCh38
rs199538197 Chr.3: 133448659-133448659 on Build GRCh38
rs934982997 Chr.3: 133448767-133448767 on Build GRCh38
rs552035038 Chr.3: 133448425-133448425 on Build GRCh38
rs575377442 Chr.3: 133448707-133448707 on Build GRCh38
rs756439680 Chr.3: 133448580-133448580 on Build GRCh38
rs376911067 Chr.3: 133448636-133448636 on Build GRCh38
rs746464807 Chr.3: 133448537-133448538 on Build GRCh38
rs751680791 Chr.3: 133448542-133448542 on Build GRCh38
rs781019303 Chr.3: 133448618-133448618 on Build GRCh38
rs768372285 Chr.3: 133448616-133448616 on Build GRCh38
rs557626440 Chr.3: 133448845-133448845 on Build GRCh38
rs772062548 Chr.3: 133448756-133448756 on Build GRCh38
rs946579746 Chr.3: 133448741-133448741 on Build GRCh38
rs768745697 Chr.3: 133448544-133448545 on Build GRCh38
rs1020176545 Chr.3: 133448851-133448851 on Build GRCh38
rs761720118 Chr.3: 133448518-133448518 on Build GRCh38
rs1028499563 Chr.3: 133448620-133448620 on Build GRCh38
rs535505104 Chr.3: 133448605-133448605 on Build GRCh38
rs200481807 Chr.3: 133448463-133448463 on Build GRCh38
rs370082928 Chr.3: 133448787-133448787 on Build GRCh38
rs767750167 Chr.3: 133448556-133448556 on Build GRCh38

View Full Product Details