Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133474882-133475391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BFSP2
Entrez Gene ID
8419
Gene Name
beaded filament structural protein 2
Gene Aliases
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN
UniGene
Hs.659862
Gene Chromosome Location
Chr.3: 133397882-133475212 on Build GRCh38
Transcripts
NM_003571.3 NR_135276.1 NR_135277.1

Gene Symbol
BFSP2-AS1
Entrez Gene ID
85003
Gene Name
BFSP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.659202
Gene Chromosome Location
Chr.3: 133426402-133491146 on Build GRCh38
Transcripts
NM_003571.3 NR_135276.1 NR_135277.1

SNP Detail

SNP ID Chromosome Location
rs986492111 Chr.3: 133475249-133475249 on Build GRCh38
rs778264861 Chr.3: 133474934-133474934 on Build GRCh38
rs933855783 Chr.3: 133475041-133475041 on Build GRCh38
rs1023514042 Chr.3: 133475145-133475145 on Build GRCh38
rs540920442 Chr.3: 133475131-133475131 on Build GRCh38
rs571904320 Chr.3: 133475160-133475160 on Build GRCh38
rs776555476 Chr.3: 133474949-133474949 on Build GRCh38
rs778064751 Chr.3: 133474966-133474966 on Build GRCh38
rs766533617 Chr.3: 133474977-133474977 on Build GRCh38
rs896890005 Chr.3: 133475110-133475110 on Build GRCh38
rs150374124 Chr.3: 133475181-133475181 on Build GRCh38
rs767743282 Chr.3: 133474995-133474995 on Build GRCh38
rs1056151656 Chr.3: 133475368-133475368 on Build GRCh38
rs1014427842 Chr.3: 133475191-133475191 on Build GRCh38
rs929704357 Chr.3: 133475358-133475358 on Build GRCh38
rs781638781 Chr.3: 133475281-133475281 on Build GRCh38
rs926039285 Chr.3: 133474950-133474950 on Build GRCh38
rs758085467 Chr.3: 133474942-133474942 on Build GRCh38
rs759824061 Chr.3: 133474994-133474994 on Build GRCh38
rs1056496690 Chr.3: 133475072-133475072 on Build GRCh38
rs753154417 Chr.3: 133475004-133475004 on Build GRCh38
rs988896660 Chr.3: 133475287-133475287 on Build GRCh38
rs765457810 Chr.3: 133474911-133474911 on Build GRCh38
rs578233592 Chr.3: 133475104-133475104 on Build GRCh38
rs1135324 Chr.3: 133475182-133475182 on Build GRCh38
rs1044231691 Chr.3: 133475039-133475039 on Build GRCh38
rs756686561 Chr.3: 133474928-133474928 on Build GRCh38
rs951118284 Chr.3: 133475241-133475241 on Build GRCh38
rs751858354 Chr.3: 133474922-133474922 on Build GRCh38
rs755802014 Chr.3: 133474912-133474912 on Build GRCh38
rs963649040 Chr.3: 133475259-133475259 on Build GRCh38
rs201953500 Chr.3: 133475008-133475008 on Build GRCh38
rs895992549 Chr.3: 133475171-133475171 on Build GRCh38
rs749884380 Chr.3: 133475018-133475018 on Build GRCh38
rs118191265 Chr.3: 133475327-133475327 on Build GRCh38
rs911123710 Chr.3: 133475319-133475319 on Build GRCh38
rs528658244 Chr.3: 133474948-133474948 on Build GRCh38
rs137944145 Chr.3: 133475324-133475324 on Build GRCh38
rs763049185 Chr.3: 133474974-133474974 on Build GRCh38
rs574253958 Chr.3: 133475198-133475198 on Build GRCh38
rs758511594 Chr.3: 133474923-133474923 on Build GRCh38
rs911176488 Chr.3: 133475051-133475051 on Build GRCh38
rs1038467164 Chr.3: 133475109-133475109 on Build GRCh38
rs78013569 Chr.3: 133475007-133475007 on Build GRCh38
rs985331335 Chr.3: 133475307-133475307 on Build GRCh38
rs757702262 Chr.3: 133475229-133475229 on Build GRCh38
rs769815007 Chr.3: 133474963-133474963 on Build GRCh38
rs886058013 Chr.3: 133475112-133475112 on Build GRCh38
rs779855632 Chr.3: 133474944-133474944 on Build GRCh38
rs139190094 Chr.3: 133475111-133475111 on Build GRCh38
rs767988370 Chr.3: 133474926-133474926 on Build GRCh38
rs1038013476 Chr.3: 133475325-133475325 on Build GRCh38
rs561545526 Chr.3: 133475296-133475296 on Build GRCh38
rs752848643 Chr.3: 133474915-133474915 on Build GRCh38
rs530264381 Chr.3: 133475258-133475261 on Build GRCh38
rs1033438559 Chr.3: 133475242-133475242 on Build GRCh38
rs537979573 Chr.3: 133475045-133475045 on Build GRCh38
rs181749711 Chr.3: 133475297-133475297 on Build GRCh38
rs774547265 Chr.3: 133474981-133474981 on Build GRCh38
rs1000185762 Chr.3: 133475237-133475237 on Build GRCh38
rs1047864826 Chr.3: 133475135-133475135 on Build GRCh38
rs541767740 Chr.3: 133475204-133475204 on Build GRCh38
rs922103561 Chr.3: 133474965-133474965 on Build GRCh38
rs376986226 Chr.3: 133475060-133475060 on Build GRCh38
rs924081752 Chr.3: 133475308-133475308 on Build GRCh38
rs979603448 Chr.3: 133475298-133475298 on Build GRCh38
rs748001305 Chr.3: 133474954-133474954 on Build GRCh38
rs753143684 Chr.3: 133474927-133474927 on Build GRCh38

View Full Product Details