Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114672815-114673327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD1
Entrez Gene ID
270
Gene Name
adenosine monophosphate deaminase 1
Gene Aliases
MAD, MADA, MMDD
UniGene
Hs.89570
Gene Chromosome Location
Chr.1: 114673098-114695618 on Build GRCh38
Transcripts
NM_000036.2 NM_001172626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599373 c.2105G>A p.G702D Chr.1: 114673253-114673253 on Build GRCh38
7355375 c.2209A>G p.N737D Chr.1: 114673149-114673149 on Build GRCh38
6599372 c.2204G>A p.G735D Chr.1: 114673253-114673253 on Build GRCh38
5353315 c.2272A>G p.M758V Chr.1: 114673185-114673185 on Build GRCh38
3385050 c.2205C>T p.G735G Chr.1: 114673252-114673252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762380006 Chr.1: 114673074-114673074 on Build GRCh38
rs6697757 Chr.1: 114673019-114673019 on Build GRCh38
rs146036570 Chr.1: 114673187-114673187 on Build GRCh38
rs142643298 Chr.1: 114673175-114673175 on Build GRCh38
rs1050131420 Chr.1: 114672944-114672944 on Build GRCh38
rs114069668 Chr.1: 114673041-114673041 on Build GRCh38
rs777802711 Chr.1: 114673176-114673176 on Build GRCh38
rs768206349 Chr.1: 114673228-114673228 on Build GRCh38
rs531229976 Chr.1: 114673251-114673251 on Build GRCh38
rs914874168 Chr.1: 114672871-114672871 on Build GRCh38
rs143596876 Chr.1: 114673212-114673212 on Build GRCh38
rs144543642 Chr.1: 114673173-114673173 on Build GRCh38
rs765157946 Chr.1: 114673087-114673089 on Build GRCh38
rs576277632 Chr.1: 114672857-114672857 on Build GRCh38
rs138460726 Chr.1: 114673155-114673155 on Build GRCh38
rs761009163 Chr.1: 114673133-114673133 on Build GRCh38
rs756744588 Chr.1: 114673065-114673065 on Build GRCh38
rs1050787988 Chr.1: 114672988-114672988 on Build GRCh38
rs764309251 Chr.1: 114673285-114673285 on Build GRCh38
rs935207162 Chr.1: 114672847-114672847 on Build GRCh38
rs773681520 Chr.1: 114673241-114673241 on Build GRCh38
rs541091632 Chr.1: 114673089-114673089 on Build GRCh38
rs759824860 Chr.1: 114673256-114673256 on Build GRCh38
rs1018620096 Chr.1: 114673156-114673156 on Build GRCh38
rs190613852 Chr.1: 114672991-114672991 on Build GRCh38
rs182475100 Chr.1: 114673013-114673013 on Build GRCh38
rs761371204 Chr.1: 114673080-114673080 on Build GRCh38
rs751538415 Chr.1: 114673286-114673286 on Build GRCh38
rs886045093 Chr.1: 114673211-114673211 on Build GRCh38
rs748789304 Chr.1: 114673226-114673226 on Build GRCh38
rs751594386 Chr.1: 114673180-114673180 on Build GRCh38
rs552905568 Chr.1: 114673275-114673275 on Build GRCh38
rs758077975 Chr.1: 114673092-114673092 on Build GRCh38
rs745489756 Chr.1: 114673188-114673188 on Build GRCh38
rs773732909 Chr.1: 114673110-114673110 on Build GRCh38
rs776996179 Chr.1: 114673252-114673252 on Build GRCh38
rs766226166 Chr.1: 114673107-114673108 on Build GRCh38
rs1054607305 Chr.1: 114672869-114672869 on Build GRCh38
rs767365369 Chr.1: 114673301-114673301 on Build GRCh38
rs945771055 Chr.1: 114672914-114672914 on Build GRCh38
rs766773123 Chr.1: 114673148-114673148 on Build GRCh38
rs778481612 Chr.1: 114673185-114673185 on Build GRCh38
rs754006954 Chr.1: 114673150-114673150 on Build GRCh38
rs377325321 Chr.1: 114673245-114673245 on Build GRCh38
rs890188851 Chr.1: 114672962-114672962 on Build GRCh38
rs891545999 Chr.1: 114672880-114672880 on Build GRCh38
rs750416342 Chr.1: 114673086-114673087 on Build GRCh38
rs571218978 Chr.1: 114673283-114673283 on Build GRCh38
rs554504528 Chr.1: 114672842-114672842 on Build GRCh38
rs570191776 Chr.1: 114673218-114673218 on Build GRCh38
rs370608479 Chr.1: 114673298-114673298 on Build GRCh38
rs752757611 Chr.1: 114673174-114673174 on Build GRCh38

View Full Product Details