Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148730088-148730589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTR1
Entrez Gene ID
185
Gene Name
angiotensin II receptor type 1
Gene Aliases
AG2S, AGTR1B, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, HAT1R
UniGene
Hs.477887
Gene Chromosome Location
Chr.3: 148697871-148743003 on Build GRCh38
Transcripts
NM_000685.4 NM_004835.4 NM_009585.3 NM_031850.3 NM_032049.3

SNP Detail

SNP ID Chromosome Location
rs991072127 Chr.3: 148730144-148730144 on Build GRCh38
rs925239709 Chr.3: 148730131-148730131 on Build GRCh38
rs574323654 Chr.3: 148730128-148730128 on Build GRCh38
rs529016869 Chr.3: 148730415-148730415 on Build GRCh38
rs143906108 Chr.3: 148730141-148730141 on Build GRCh38
rs915773074 Chr.3: 148730172-148730172 on Build GRCh38
rs75367686 Chr.3: 148730277-148730277 on Build GRCh38
rs919968878 Chr.3: 148730123-148730123 on Build GRCh38
rs534066472 Chr.3: 148730347-148730347 on Build GRCh38
rs552746485 Chr.3: 148730431-148730431 on Build GRCh38
rs550710476 Chr.3: 148730297-148730297 on Build GRCh38
rs936459917 Chr.3: 148730232-148730232 on Build GRCh38
rs939727079 Chr.3: 148730235-148730235 on Build GRCh38
rs908338991 Chr.3: 148730545-148730545 on Build GRCh38
rs1014777871 Chr.3: 148730364-148730364 on Build GRCh38
rs147283545 Chr.3: 148730146-148730146 on Build GRCh38
rs115123914 Chr.3: 148730309-148730309 on Build GRCh38
rs1038049278 Chr.3: 148730269-148730269 on Build GRCh38
rs530888946 Chr.3: 148730290-148730290 on Build GRCh38
rs528800862 Chr.3: 148730147-148730147 on Build GRCh38
rs963524842 Chr.3: 148730374-148730374 on Build GRCh38
rs56383020 Chr.3: 148730543-148730543 on Build GRCh38
rs545348919 Chr.3: 148730247-148730247 on Build GRCh38
rs991382194 Chr.3: 148730166-148730166 on Build GRCh38
rs908288868 Chr.3: 148730228-148730228 on Build GRCh38
rs1005178032 Chr.3: 148730357-148730357 on Build GRCh38
rs949606428 Chr.3: 148730165-148730167 on Build GRCh38
rs190344670 Chr.3: 148730469-148730469 on Build GRCh38
rs990789128 Chr.3: 148730460-148730460 on Build GRCh38
rs1001326892 Chr.3: 148730398-148730398 on Build GRCh38
rs565937018 Chr.3: 148730562-148730562 on Build GRCh38
rs924083965 Chr.3: 148730224-148730224 on Build GRCh38
rs971262054 Chr.3: 148730479-148730479 on Build GRCh38
rs971023074 Chr.3: 148730494-148730494 on Build GRCh38
rs991434646 Chr.3: 148730454-148730454 on Build GRCh38
rs778629974 Chr.3: 148730299-148730299 on Build GRCh38
rs967151116 Chr.3: 148730122-148730122 on Build GRCh38
rs959619497 Chr.3: 148730436-148730436 on Build GRCh38
rs902602912 Chr.3: 148730384-148730384 on Build GRCh38
rs981225093 Chr.3: 148730219-148730219 on Build GRCh38
rs949853207 Chr.3: 148730197-148730197 on Build GRCh38
rs932940909 Chr.3: 148730285-148730285 on Build GRCh38
rs750673785 Chr.3: 148730354-148730354 on Build GRCh38

View Full Product Details