Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149779997-149780532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKUB1
Entrez Gene ID
389161
Gene Name
ankyrin repeat and ubiquitin domain containing 1
Gene Aliases
C3orf16
UniGene
Hs.665988
Gene Chromosome Location
Chr.3: 149760648-149792823 on Build GRCh38
Transcripts
NM_001144960.2 NM_001315505.1 NM_001315506.1 XM_011512798.1 XM_011512799.1 XM_011512801.1 XM_011512802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1039806 c.369C>T p.P123P Chr.3: 149780321-149780321 on Build GRCh38
3002879 c.399G>T p.R133S Chr.3: 149780291-149780291 on Build GRCh38
1039807 c.261G>A p.V87V Chr.3: 149780429-149780429 on Build GRCh38
5995219 c.399G>A p.R133R Chr.3: 149780291-149780291 on Build GRCh38
5032018 c.274A>T p.T92S Chr.3: 149780416-149780416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746634770 Chr.3: 149780439-149780439 on Build GRCh38
rs750257988 Chr.3: 149780321-149780321 on Build GRCh38
rs749644987 Chr.3: 149780284-149780284 on Build GRCh38
rs374221123 Chr.3: 149780494-149780494 on Build GRCh38
rs377158470 Chr.3: 149780434-149780434 on Build GRCh38
rs764978334 Chr.3: 149780196-149780196 on Build GRCh38
rs188852314 Chr.3: 149780063-149780063 on Build GRCh38
rs769789290 Chr.3: 149780450-149780450 on Build GRCh38
rs60894864 Chr.3: 149780302-149780302 on Build GRCh38
rs762748277 Chr.3: 149780474-149780474 on Build GRCh38
rs7622440 Chr.3: 149780035-149780035 on Build GRCh38
rs928862514 Chr.3: 149780334-149780334 on Build GRCh38
rs775771836 Chr.3: 149780112-149780112 on Build GRCh38
rs980262945 Chr.3: 149780504-149780504 on Build GRCh38
rs751746279 Chr.3: 149780349-149780349 on Build GRCh38
rs577775162 Chr.3: 149780128-149780128 on Build GRCh38
rs1002537688 Chr.3: 149780352-149780352 on Build GRCh38
rs958016487 Chr.3: 149780295-149780295 on Build GRCh38
rs138748319 Chr.3: 149780193-149780193 on Build GRCh38
rs778423400 Chr.3: 149780421-149780421 on Build GRCh38
rs770084022 Chr.3: 149780279-149780279 on Build GRCh38
rs771667167 Chr.3: 149780295-149780295 on Build GRCh38
rs893575751 Chr.3: 149780490-149780490 on Build GRCh38
rs1030595759 Chr.3: 149780477-149780477 on Build GRCh38
rs972710688 Chr.3: 149780344-149780344 on Build GRCh38
rs749733005 Chr.3: 149780431-149780431 on Build GRCh38
rs566910040 Chr.3: 149780506-149780506 on Build GRCh38
rs763999128 Chr.3: 149780178-149780178 on Build GRCh38
rs1037150589 Chr.3: 149780084-149780084 on Build GRCh38
rs753162142 Chr.3: 149780367-149780367 on Build GRCh38
rs1011033493 Chr.3: 149780491-149780491 on Build GRCh38
rs938703683 Chr.3: 149780343-149780343 on Build GRCh38
rs199517574 Chr.3: 149780432-149780432 on Build GRCh38
rs376831846 Chr.3: 149780270-149780270 on Build GRCh38
rs942542277 Chr.3: 149780419-149780419 on Build GRCh38
rs181647061 Chr.3: 149780267-149780267 on Build GRCh38
rs769038319 Chr.3: 149780217-149780217 on Build GRCh38
rs944108312 Chr.3: 149780499-149780499 on Build GRCh38
rs761761944 Chr.3: 149780318-149780318 on Build GRCh38
rs200616244 Chr.3: 149780320-149780320 on Build GRCh38
rs965050645 Chr.3: 149780226-149780226 on Build GRCh38
rs542297707 Chr.3: 149780209-149780209 on Build GRCh38
rs755071802 Chr.3: 149780243-149780243 on Build GRCh38
rs948957247 Chr.3: 149780028-149780028 on Build GRCh38
rs370102552 Chr.3: 149780484-149780484 on Build GRCh38
rs62622807 Chr.3: 149780310-149780310 on Build GRCh38
rs771922544 Chr.3: 149780294-149780294 on Build GRCh38
rs76835165 Chr.3: 149780462-149780462 on Build GRCh38
rs16862241 Chr.3: 149780360-149780360 on Build GRCh38
rs776254788 Chr.3: 149780442-149780442 on Build GRCh38
rs34140953 Chr.3: 149780456-149780456 on Build GRCh38
rs980316724 Chr.3: 149780327-149780327 on Build GRCh38
rs374411928 Chr.3: 149780394-149780394 on Build GRCh38
rs35021401 Chr.3: 149780368-149780368 on Build GRCh38
rs747580476 Chr.3: 149780443-149780443 on Build GRCh38
rs369413252 Chr.3: 149780280-149780280 on Build GRCh38
rs533629917 Chr.3: 149780472-149780472 on Build GRCh38
rs776950688 Chr.3: 149780292-149780292 on Build GRCh38
rs865810437 Chr.3: 149780290-149780290 on Build GRCh38
rs760119759 Chr.3: 149780306-149780306 on Build GRCh38
rs772702129 Chr.3: 149780072-149780072 on Build GRCh38
rs77152758 Chr.3: 149780345-149780345 on Build GRCh38
rs141785874 Chr.3: 149780194-149780194 on Build GRCh38
rs73870412 Chr.3: 149780070-149780070 on Build GRCh38
rs558072191 Chr.3: 149780111-149780111 on Build GRCh38
rs758668980 Chr.3: 149780216-149780216 on Build GRCh38
rs768075066 Chr.3: 149780441-149780441 on Build GRCh38
rs1021803250 Chr.3: 149780493-149780493 on Build GRCh38
rs144845141 Chr.3: 149780130-149780130 on Build GRCh38
rs766644191 Chr.3: 149780347-149780347 on Build GRCh38
rs889721726 Chr.3: 149780149-149780149 on Build GRCh38
rs1044421313 Chr.3: 149780029-149780029 on Build GRCh38
rs751656851 Chr.3: 149780241-149780241 on Build GRCh38
rs768148102 Chr.3: 149780307-149780307 on Build GRCh38
rs186123914 Chr.3: 149780359-149780359 on Build GRCh38

View Full Product Details