The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033025.2 | NP_001028197.1 | 2 - 3 | 622 | 97 |
NM_001261440.1 | NP_001248369.1 | 2 - 3 | 622 | 97 | |
NM_001261442.1 | NP_001248371.1 | 2 - 3 | 658 | 97 | |
NM_001439.3 | NP_001430.1 | 2 - 3 | 658 | 97 | |
XM_005270621.1 | XP_005270678.1 | 3 - 4 | 488 | 97 | |
XM_017000651.1 | XP_016856140.1 | 2 - 3 | 262 | 97 | |
GenBank mRNA | AB009284.1 | - | 2 - 3 | 291 | 97 |
AF000416.1 | - | 2 - 3 | 291 | 97 | |
AK096914.1 | - | 2 - 3 | 255 | 97 | |
AK312895.1 | - | 2 - 3 | 227 | 97 | |
BC014910.1 | - | 2 - 3 | 178 | 97 | |
BC036015.1 | - | 2 - 3 | 268 | 97 | |
BC045681.1 | - | 2 - 3 | 1441 | 97 | |
BX103291.1 | - | 2 - 3 | 291 | 97 |